Sterile & PCR ready
Consumables and supplies fit for the rigors of the most sensitive PCR-based science
Look for the Thermo Scientific Sterile & PCR ready product designation when buying consumables for your most sensitive PCR based studies. Products with this designation are among the highest quality available and most comprehensively tested to detect impurities. The Thermo Scientific Sterile & PCR ready designation includes combination testing for sterility and elimination of endotoxins (see Sterile designation), as well as testing for the elimination of RNase, DNase, human DNA, and PCR inhibitors (see PCR ready designation). Products that meet this dual designation are intended to support the extreme demands of PCR-based science where sample material is limited or especially valuable and in applications where any contamination poses enhanced risk to results. |
To receive the Sterile & PCR ready designation, products must meet the following criteria:
✓ Minimum Sterility Assurance Level (SAL) with ISO 11137
✓ Endotoxin (pyrogen) free
✓ Human DNA free
✓ RNase Free
✓ DNase Free
✓ PCR inhibitor free
Additional criteria met:
✓ Made from high quality resins and raw materials
✓ Manufactured in accordance with ISO 9001 and 13485 standards
✓ Low wetting for accurate liquid measurements
✓ High chemical resistance for durability in most routine lab work
✓ High thermal resistance for stability under wide range of temperatures
✓ High centrifugation stability*
✓ High transparency for good visual sample monitoring*
✓ Precisely shaped for outstanding function and critical measurements
*For applicable products only
Shop Sterile & PCR ready products
Thermo Scientific Sterile & PCR ready designated products are recommended for use with the following application areas and techniques:
Application areas
→ Environmental monitoring
→ Food and beverage quality control
→ Forensic human identification
→ Medical research
→ Microbiology
→ Molecular biology
→ Stem cell research
→ Transgenic plant/animal studies
→ Water quality control
Techniques
→ Bacterial and yeast cultures
→ Cell and tissue cultures
→ DNA analysis (PCR, restriction analysis, hybridization, sequencing, NGS)
→ Isolation and storage of RNA
→ Isolation and storage of DNA
→ Mitochondrial DNA analysis
→ RNA analysis
Browse popular products certified under our Thermo Scientific Sterile & PCR ready designation.
- ART Barrier Reload System ›
- ART Barrier Extended Length Pipette Tips ›
- ART Barrier Pipette Tips in Hinged Racks ›
- ART Barrier Speciality Pipette Tips ›
- ART Barrier Low-Retention Pipette Tips ›
- ART Barrier Reload Insert Pipette Tips ›
- ART Barrier Pipette Tips in Lift-off Lid Rack ›
- ART Wide Bore Filtered Pipette Tips ›
- ART Gel Loading Pipette Tips ›
- ART Barrier Reload Insert, Extended Length Pipette Tips ›
- ART Barrier Extended Length Reload System ›
- ART 1000 REACH RACKED barrier sterile pipette tips ›
- Extended Length 200μL ART Barrier Speciality Filter Pipettor Tip ›
- ART Barrier Specialty Pipette Tips, 200 XLG ›
- Automation Tips for the Eppendorf epMotion Automated Pipetting System ›
- Automation Tips for Caliper Zymark Liquid Handling Workstations ›
- Automation Tips for PerkinElmer/Packard Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads ›
- Automation Tips for Tecan Liquid Handling Platforms ›
- Automation Tips for Tecan Liquid Handling Platforms with Te-MO 96 Multichannel Arm ›
- Automation Tips for Beckman Liquid Handling Systems with AP96 Dispense Head ›
- Automation Tips for Beckman Liquid Handling Systems with MP20 and MP200 Dispense Heads ›
- Conductive Automation Tips for Qiagen Workstations ›
- Automation Tips for Qiagen Workstations ›
**Sterile & PCR ready Fit for Purpose Limits of Detection apply to Finntip System Tips produced beginning with lot 22258XX.
- Matrix 2D Barcoded 12mL Screw-top Storage Tubes ›
- Matrix Blank and Alphanumeric Storage Tubes ›
- Matrix 1.0mL ScrewTop Tubes ›
- Matrix 2D Barcoded Open-Top Storage Tubes ›
- Matrix 2D Barcoded Low Volume ScrewTop Storage Tubes ›
- Matrix 1.0mL ScrewTop Tubes in Barcoded Latch Racks ›
- Matrix 0.5mL ScrewTop Tubes in Barcoded Latch Racks ›
- Matrix ScrewTop Tube Caps, Cap Trays and Cap Inserts ›
- Matrix 500μL ScrewTop Tubes ›
- Matrix ScrewTop Tri-coded Tubes ›