Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips
Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips
Thermo Scientific™

Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Unit Size
2160P200 μLCase of 3072
2160PPK
also known as 2160P
200 μLPack of 768
Catalog number 2160P
Price (USD)/ Case of 3072
467.00
Online exclusive
888.00 
Save 421.00 (47%)
-
Add to cart
Volume (Metric):
200 μL
Unit Size:
Case of 3072
Price (USD)/ Case of 3072
467.00
Online exclusive
888.00 
Save 421.00 (47%)
Add to cart
Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips
Catalog number2160P
Price (USD)/ Case of 3072
467.00
Online exclusive
888.00 
Save 421.00 (47%)
-
Add to cart
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetting for a variety of applications:

Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ ™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
DescriptionART XLP 200, Filtered, Sterile,
Volume (Metric)200 μL
FilteredFiltered
SterilitySterile
ColorNatural
ModelART XLP 200
Length (English)3.59 in.
Length (Metric)9.12cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatRack
Tip StyleExtended Length
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 3072