ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
Thermo Scientific™

ClipTip™ Filtered 384-Format Pipette Tips

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip Style
94420153125 μLStandard 384
9442007312.5 μLExtended Length
9442005312.5 μLStandard 384
9442010330 μLStandard 384
Catalog number 94420153
Price (USD)/ Case of 3840
562.50
Online exclusive
630.00 
Save 67.50 (11%)
-
Add to cart
Volume (Metric):
125 μL
Tip Style:
Standard 384
Price (USD)/ Case of 3840
562.50
Online exclusive
630.00 
Save 67.50 (11%)
Add to cart
ClipTip™ Filtered 384-Format Pipette Tips
Catalog number94420153
Price (USD)/ Case of 3840
562.50
Online exclusive
630.00 
Save 67.50 (11%)
-
Add to cart

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes. Featuring an innovativesnap and seal tip attachment mechanism, the pipette tips ensure a light force with a secure tip attachment, enabling greater efficiency and confidence in your work.

Key Features:

  • All ClipTip 384-format tips feature a ‘Snap and Seal’ tip attachment mechanism that ensures light force and secure tip attachment
  • Each tip has incorporated small protrusions that snap on the tip fitting flange of the pipette
  • Ideal for 384-format microplate applications
  • Filtered tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations

Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Versatile Tip Range

  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended lenght tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables

Immediate traceability

  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert

Note:

Availability may vary by country.

Specifications
DescriptionClipTip 384 125, Filtered, Sterile, Rack
Volume (Metric)125 μL
FilteredFiltered
SterilitySterile
AutoclavableNot Autoclavable
Length (English)2.52 in.
Length (Metric)6.4cm
FormatRack
Tip StyleStandard 384
Packaging10 x 384 tips/rack
Color CodeYellow
For Use With (Equipment)384-Format Microplate, E1-ClipTip Electronic
Product LineClipTip
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 3840