Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredFor Use With (Equipment)Fit for PurposeUnit Size
919-021130 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyCase of 4800
912-0218 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyCase of 4800
912-26110 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterileCase of 4800
912-261PK
also known as 912-261
10 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterilePack of 960
912-26210 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
912-262PK
also known as 912-262
10 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
916-02140 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyCase of 4800
916-021PK
also known as 916-021
40 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyPack of 960
916-26150 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterileCase of 4800
916-261PK
also known as 916-261
50 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterilePack of 960
916-26250 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
916-262PK
also known as 916-262
50 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
916-262-0550 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
916-262-05PK
also known as 916-262-05
50 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
918-02120 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyCase of 4800
918-021PK
also known as 918-021
20 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyPack of 960
918-021-0520 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyCase of 4800
918-021-05PK
also known as 918-021-05
20 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyPack of 960
918-26120 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterileCase of 4800
918-261PK
also known as 918-261
20 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterilePack of 960
918-261-0520 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterileCase of 4800
918-26220 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
918-262PK
also known as 918-262
20 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
918-262-0520 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
918-262-05PK
also known as 918-262-05
20 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
919-021PK
also known as 919-021
130 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyPack of 960
919-021-05130 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyCase of 4800
919-021-05PK
also known as 919-021-05
130 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyPack of 960
919-021G130 μLFilteredBeckman Coulter Liquid Handling SystemsSterile & PCR ReadyCase of 4800
919-261250 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterileCase of 4800
919-261PK
also known as 919-261
250 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterilePack of 960
919-261-05250 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterileCase of 4800
919-261-05PK
also known as 919-261-05
250 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterilePack of 960
919-261G250 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterileCase of 4800
919-261GPK
also known as 919-261G
250 μLNon-filteredBeckman Coulter Liquid Handling SystemsSterilePack of 960
919-262250 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
919-262PK
also known as 919-262
250 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
919-262-05250 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
919-262-05PK
also known as 919-262-05
250 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
919-262G250 μLNon-filteredBeckman Coulter Biomek FXCertified QualityCase of 4800
919-262GPK
also known as 919-262G
250 μLNon-filteredBeckman Coulter Biomek FXCertified QualityPack of 960
276050 μLFilteredUniversal Fit PipettorSterileCase of 4800
Catalog number 919-021
Price (USD)/ Case of 4800
614.00
Online exclusive
1060.00 
Save 446.00 (42%)
-
Add to cart
Volume (Metric):
130 μL
Filtered:
Filtered
For Use With (Equipment):
Beckman Coulter Liquid Handling Systems
Fit for Purpose:
Sterile & PCR Ready
Unit Size:
Case of 4800
Price (USD)/ Case of 4800
614.00
Online exclusive
1060.00 
Save 446.00 (42%)
Add to cart
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Catalog number919-021
Price (USD)/ Case of 4800
614.00
Online exclusive
1060.00 
Save 446.00 (42%)
-
Add to cart
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
Volume (Metric)130 μL
FilteredFiltered
SterilitySterile
ColorClear
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
FormatRack
Tip StyleAutomation
Packaging96 tips/rack, 10 racks/pack, 5 packs/case
Quantity4800 tips
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
Product LineBeckman, Beckman
Fit for PurposeSterile & PCR Ready
Unit SizeCase of 4800