Find the right rotor for your SpeedVac
Easy-to-use circular rotors have different tube-hole sizes to accommodate a variety of sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
Supported models
These rotors are compatible with SPD1030, SDP120, SPD130, SPD140DDA
Supported legacy systems: SPD1010, AES1010, AES1000, SPD111V, SPD121P, SPD131DDA, SC110A
Format | Capacity | Rotor part # | Rotor system |
---|---|---|---|
Microtubes | |||
1.5mL | 40 | RD40-11 | Traditional |
1.5mL | 64 | RH64-11 | Traditional |
1.5mL | 120 | RH120-11 | Traditional |
Glass & plastic tubes | |||
6x50mm | 100 | RH100-6 | Traditional |
8x29mm | 100 | RH100-8 | Traditional |
12x75mm | 40 | RH40-12 | Traditional |
12x75mm | 72 | RH72-12 | Traditional |
13x100mm | 32 | RH32-13 | Traditional |
17.5x102mm, 15mL Corex® | 8 | RH8-17.5 | Traditional |
18x100, 17x95, 16x100mm | 8 | RH8-18 | Traditional |
Centrifuge tubes | |||
15mL conical | 10 | RH10-15 | Traditional |
50mL conical | 6 | RH6-50 | Traditional |
Flasks | |||
100mL pear shaper | 4 | RH4-100 | Traditional |
Vials | |||
12x32mm | 60 | RH60-12-40 | Traditional |
12x40mm | 24 | RH24-15 | Traditional |
20x47mm | 60 | RH60-12-40 | Traditional |
20x60mm | 12 | RH12-20 | Traditional |
18x52mm mini-scintillation | 12 | RH12-20 | Traditional |
28x60mm scintillation | 24 | RH24-18 | Traditional |
Microwell plates | |||
Shallow well plates | 6 | RHSW6MP* | Traditional |
Shallow well plates | 2 | RH2MP | Traditional |
Deep well plates | 2 | RHDW2MP* | Traditional |
*Not available for SPD1030 models
Supported models
These rotors are compatible with SPD1030, SDP120, SPD130, SPD140DDA
Supported legacy systems: SPD1010, AES1010, AES1000, SPD111V, SPD121P, SPD131DDA, SC110A
Format | Capacity | Rotor part # | Rotor system |
---|---|---|---|
Microtubes | |||
1.5mL | 40 | RD40-11 | Traditional |
1.5mL | 64 | RH64-11 | Traditional |
1.5mL | 120 | RH120-11 | Traditional |
Glass & plastic tubes | |||
6x50mm | 100 | RH100-6 | Traditional |
8x29mm | 100 | RH100-8 | Traditional |
12x75mm | 40 | RH40-12 | Traditional |
12x75mm | 72 | RH72-12 | Traditional |
13x100mm | 32 | RH32-13 | Traditional |
17.5x102mm, 15mL Corex® | 8 | RH8-17.5 | Traditional |
18x100, 17x95, 16x100mm | 8 | RH8-18 | Traditional |
Centrifuge tubes | |||
15mL conical | 10 | RH10-15 | Traditional |
50mL conical | 6 | RH6-50 | Traditional |
Flasks | |||
100mL pear shaper | 4 | RH4-100 | Traditional |
Vials | |||
12x32mm | 60 | RH60-12-40 | Traditional |
12x40mm | 24 | RH24-15 | Traditional |
20x47mm | 60 | RH60-12-40 | Traditional |
20x60mm | 12 | RH12-20 | Traditional |
18x52mm mini-scintillation | 12 | RH12-20 | Traditional |
28x60mm scintillation | 24 | RH24-18 | Traditional |
Microwell plates | |||
Shallow well plates | 6 | RHSW6MP* | Traditional |
Shallow well plates | 2 | RH2MP | Traditional |
Deep well plates | 2 | RHDW2MP* | Traditional |
*Not available for SPD1030 models