Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Have Questions?
Change view
Catalog Number
Holds
RD36
36
RD24
24
RD2MP
2 Shallow-well Plates
RD48
48
RD72
72
Catalog number RD36
Price (USD)/ Each
-
Request A Quote
Holds:
36
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.