Find the right Rotor for your SpeedVac
SpeedVac rotors for large capacity concentrators for use with Savant™ SpeedVac™ Vacuum Concentrators.
Supported models
These rotors are compatible with SPD210, SPD2030, SPD300DDA (requires RAD200)
Supported legacy systems: SC210A, SPD2010, AES2010, AES1000, SC250EXP, SC250DDA
Format | Capacity | Rotor or block part # | Rotor system |
---|---|---|---|
Microtubes | |||
1.5mL | 50 | RBA25-11-39 | Set of 2 blocks for PRO |
1.5mL | 100 | RBA25-11-39 (qty 2) | Set of 3 blocks for PRO |
1.5mL | 200 | RH200-12* | Traditional |
Glass & plastic tubes | |||
6x32mm | 252 | RBA126-6-32 | Set of 2 blocks for PRO |
6x32mm | 504 | RBA126-6-32 (qty 2) | Set of 2 blocks for PRO |
12x75mm | 200 | RH200-12* | Traditional |
13x100mm | 70 | RBA35-13-100 | Set of 2 blocks for PRO |
13x100mm | 118 | RH200-12* | Traditional |
13x100mm | 140 | RBA35-13-100 (qty 2) | Set of 2 blocks for PRO |
16x100mm | 48 | RBA24-16-100 | Set of 2 blocks for PRO |
16x100mm | 96 | RBA24-16-100 (qty 2) | Set of 2 blocks for PRO |
16x125mm | 48 | RH48-18-125* | Traditional |
16x125mm | 30 | RBA15-16-125 | Set of 2 blocks for PRO |
17x60mm | 48 | RBA24-16-125 | Set of 2 blocks for PRO |
17x60mm | 96 | RBA24-16-125 (qty 2) | Set of 2 blocks for PRO |
16x125mm | 60 | RBA15-16-125 (qty 2) | Set of 2 blocks for PRO |
18x125mm | 48 | RH48-18-125* | Traditional |
18x150, 16x125mm | 32 | RH32-18-150 | Traditional |
28x150mm | 12 | RH12-29** | Traditional |
Centrifuge tubes | |||
15mL conical | 30 | RH60-17-100* | Traditional |
15mL conical | 52 | RH52-15* | Traditional |
50mL conical | 12 | RH12-29** | Traditional |
Flasks | |||
100mL round bottom | 2 | RBA2-FLSK-100** | Traditional |
100mL pear shaped | 8 | RH8-200** | Traditional |
250mL round bottom | 1 | RBA1-FLSK-250** | Traditional |
Vials | |||
1 dram (15x45mm), 4mL | 192 | RH192-15* | Traditional |
28x58mm, 20mL scintillation | 24 | RBA12-28-58 | Set of 2 blocks for PRO |
28x58mm, 20mL scintillation | 48 | RBA12-28-58 (qty 2) | Set of 2 blocks for PRO |
28x60mm scintillation | 50 | RH50-28-60* | Traditional |
19x65mm flat bottom | 48 | RBA24-19-65 | Set of 2 blocks for PRO |
19x65mm flat bottom | 96 | RBA24-19-65 (qty 2) | Set of 2 blocks for PRO |
Microwell plates | |||
Shallow well plates | 12 | MPTR12-210* | Traditional |
Shallow well plates | 4 | UPC-1 | Set of 4 carriers for PRO |
Shallow well plates | 8 | UPC-2 | Set of 4 carriers for PRO |
Shallow well plates | 20 | UPC-5 | Set of 4 carriers for PRO |
Deep well plates | 4 | UPC-1 | Set of 4 carriers for PRO |
Deep well plates | 8 | UPC-2 | Set of 4 carriers for PRO |
Deep well plates | 8 | MPTR8-210* | Traditional |
*SPD300DDA requires RAD200 to utilize this format
**Format not supported by SPD300DDA