PRO Rotor System
An alternative to traditional rotors is our PRO rotor system. This is a highly versatile rotor system consists of a four-position rotor along with mix-and-match carriers and blocks. This system can be used with the following Savant SpeedVac vacuum concentrator models: SPD210, SPD2030, and SPD300. Use the charts below to identify the necessary system or configure your PRO rotor system using individual components. Unless otherwise indicated, all rotors, blocks, and carriers are sold separately.
PRO Rotor Systems
A complete system to upgrade your current SpeedVac rotors.
Image | Catalog# | Description | No. of Tubes/Plates |
---|---|---|---|
PRO-10 | Includes single-shelf, heavy-duty rotor kit, consisting of UPR-4A rotor, UPC-1 carrier set and SUMAX400 upper magnetic assembly | See rotor block table | |
PRO-20 | Includes single-shelf, heavy-duty rotor kit, consisting of UPR-4A rotor, UPC-2 carrier set and SUMAX400 upper magnetic assembly | 8 deep-well plates | |
PRO-50 | Includes single-shelf, heavy-duty rotor kit, consisting of UPR-4A rotor, UPC-5 carrier set and SUMAX400 upper magnetic assembly | 20 shallow-well plates |
PRO Rotor Components
These blocks are compatible with the UPC-1 carrier in our PRO rotor system and expand the types of tubes/vessels you can use in your SpeedVac System.
Cat No. | Description | No. of Tubes |
---|---|---|
RBA12-25-52 | 24 X 52mm, 2 dram vials | 12 |
RBA12-27-60 | 27 X 60mm, 5 dram vials | 12 |
RBA12-28-58 | 28 X 58mm, 20ml scintillation vials | 12 |
RBA15-16-125 | 16 X 125mm tubes | 15 |
RBA24-15-45 | 15 X 45mm, 1 dram vials | 24 |
RBA24-16-100 | 16 X 100mm tubes | 24 |
RBA24-17-31 | 17 X 31mm, 3mLvials | 24 |
RBA24-19-65 | 19 X 65mm flat bottom vials | 24 |
RBA35-13-100 | 13 X 100mm tubes | 35 |
RBA126-6-32 | 6 X 32mm tubes | 126 |
RBA20-12-125 | 12 X 125mm tubes | 20 |
RBA24-17-60 | 17 X 60mm tubes | 24 |
RBA25-11-39 | 1.5mL micro-centrifuge tubes | 25 |
RBA30-8-30 | 0.5mL micro-centrifuge tubes | 30 |
RBA6-28-110 | 28 X 110mm tubes | 6 |
RBA1-FLSK-250 | Flasks: 1 x 250mL round bottom, Large capacity models | 1 bottle flask |
RBA2-FLSK-100 | Flasks: 2 x 100mL round bottom, Large capacity models | 2 bottle flasks |
RBSH | Rotor block adapter for Array-It Side Holders (Microarrays), set of 2 | 27 slides |
UPC carriers drop into our UPR-4A rotor and can support blocks for tubes and slides or deep and shallow-well plates.
Cat No. | Description | Shallow-well Microplates | Deep-well Plates | Rotor Blocks for Vials and Tubes |
---|---|---|---|---|
UPC-1 | Single-shelf, set of 4 carriers | 4 | 4 | 4 |
UPC-2 | Double-shelf, set of 4 carriers | 8 | 8 | - |
UPC-5 | Five-shelf, set of 4 carriers | 20 | - | - |
Thermo Scientific FPR-4A 4-position swing-out horizontal universal plate rotor for UPC carriers
Thermo Scientific SUMAX400 Magnetic Assembly
These blocks are compatible with the UPC-1 carrier in our PRO rotor system and expand the types of tubes/vessels you can use in your SpeedVac System.
Cat No. | Description | No. of Tubes |
---|---|---|
RBA12-25-52 | 24 X 52mm, 2 dram vials | 12 |
RBA12-27-60 | 27 X 60mm, 5 dram vials | 12 |
RBA12-28-58 | 28 X 58mm, 20ml scintillation vials | 12 |
RBA15-16-125 | 16 X 125mm tubes | 15 |
RBA24-15-45 | 15 X 45mm, 1 dram vials | 24 |
RBA24-16-100 | 16 X 100mm tubes | 24 |
RBA24-17-31 | 17 X 31mm, 3mLvials | 24 |
RBA24-19-65 | 19 X 65mm flat bottom vials | 24 |
RBA35-13-100 | 13 X 100mm tubes | 35 |
RBA126-6-32 | 6 X 32mm tubes | 126 |
RBA20-12-125 | 12 X 125mm tubes | 20 |
RBA24-17-60 | 17 X 60mm tubes | 24 |
RBA25-11-39 | 1.5mL micro-centrifuge tubes | 25 |
RBA30-8-30 | 0.5mL micro-centrifuge tubes | 30 |
RBA6-28-110 | 28 X 110mm tubes | 6 |
RBA1-FLSK-250 | Flasks: 1 x 250mL round bottom, Large capacity models | 1 bottle flask |
RBA2-FLSK-100 | Flasks: 2 x 100mL round bottom, Large capacity models | 2 bottle flasks |
RBSH | Rotor block adapter for Array-It Side Holders (Microarrays), set of 2 | 27 slides |
UPC carriers drop into our UPR-4A rotor and can support blocks for tubes and slides or deep and shallow-well plates.
Cat No. | Description | Shallow-well Microplates | Deep-well Plates | Rotor Blocks for Vials and Tubes |
---|---|---|---|---|
UPC-1 | Single-shelf, set of 4 carriers | 4 | 4 | 4 |
UPC-2 | Double-shelf, set of 4 carriers | 8 | 8 | - |
UPC-5 | Five-shelf, set of 4 carriers | 20 | - | - |
Thermo Scientific FPR-4A 4-position swing-out horizontal universal plate rotor for UPC carriers
Thermo Scientific SUMAX400 Magnetic Assembly