CaptureSelect conjugated ligands are available for use in a range of analytical assays and can be used for the specific detection, quantitation, and characterization of antibodies, antibody fragments, proteins, hormones, and viral vectors. The affinity ligands are chemically conjugated to either biotin, HRP, or a fluorescent dye.
The biotinylated ligands are conjugated using an appropriate spacer that retains binding reactivity of the ligand when immobilized onto streptavidin-functionalized surfaces, thereby allowing for use in activation and kinetic assays.
Contact us Download application guide
CaptureSelect technology
Conventional antibodies, such as immunoglobin G (IgG), are composed of a heavy chain and a light chain. In some species, the IgGs lack the light chain and are composed solely of the heavy chain, yielding a heavy-chain IgG (hc-IgG). CaptureSelect affinity ligands are created by a proprietary technology based on recombinant single domain antibodies. The ligand is a ~14 kDa single domain fragment (VHH) containing the three complementary determining regions (CDRs) that form the antigen binding domain (Figure 1).
To generate the VHH ligand, the heavy-chain antibodies are pared down to focus on only those parts that are important for binding an antigen, which includes the CDR. The VHH ligands maintain high affinity and specific antigen binding while being one-tenth the size of traditional antibodies. The VHH ligands are recombinantly expressed in the yeast Saccharomyces cerevisiae in a production process free of any animal components (Animal Origin-Free).
Figure 1. A regular antibody (IgG) compared to a heavy-chain antibody (hc-Ig) lacking the light chains. CaptureSelect ligands are based on the single domain fragment (VHH) from these hc-IgGs. On the right side, a ribbon diagram of a VHH ligand is shown. The CDRs are indicated in red, green, and blue.
Ligand applications
CaptureSelect conjugated ligands can be used in multiple areas, such as discovery and screening, detection, quantitation, and other product or process analytics.
Applications can include:
For examples, protocols, and publications highlighting the conjugated ligands, download the application guide or watch the webinar on the versatility of single domain antibody technology.
Figure 2. Representation of a CaptureSelect biotinylated ligand bound to its target protein. The biotin molecules are chemically conjugated to the affinity ligand through a spacer, allowing immobilization on streptavidin-functionalized surfaces.
Webinar: Demonstrating versatility of single domain antibody technology in the development of antibody therapeutics
Watch this webinar to discover:
Watch now ›
CaptureSelect conjugated affinity ligands
We have created an extensive list of ligands directed against:
You can find more detailed information on the selectivity and use of these products in the applications guide or by viewing the specific product pages linked below.
CaptureSelect conjugated ligands for antibodies and antibody fragments
CaptureSelect ligand | Biotin | Biotin |
100 µg | 500 µg | |
Anti-IgG-Fc (human) | 7103262100 | 7103262500 |
Anti-IgG-CH1 | 7103202100 | 7103202500 |
Anti-LC-Kappa(human) | 7103272100 | 7103272500 |
Anti-LC-Lambda(human) | 7103082100 | 7103082500 |
Anti-Free LC-Kappa(human) | 7103292100 | 7103292500 |
Anti-IgG3 | 7103042100 | 7103042500 |
Anti-IgG4 | 7102902100 | 7102902500 |
Anti-IgA | 7102882100 | 7102882500 |
Anti-IgM | 7102892100 | 7102892500 |
Anti-IgE | 7103542100 | 7103542500 |
Anti-IgG-FC(multi-species) | 7102852100 | 7102852500 |
Anti-LC-Kappa(murine) | 7103152100 | 7103152500 |
Anti-LC-Lambda(mouse) | 7103232100 | 7103232500 |
Anti-IgG-FC (Rabbit) | 7103642100 | 7103642500 |
CaptureSelect conjugated ligands for proteins and hormones
CaptureSelect ligand | Biotin | Biotin |
100 µg | 500 µg | |
Anti-Factor VII | 7102992100 | 7102992500 |
Anti-Factor VIII | 7102862100 | 7102862500 |
Anti-Factor IX | 7103002100 | 7103002500 |
Anti-Factor X | 7103702100 | 7103702500 |
Anti-AAT | 7102872100 | 7102872500 |
Anti-Fibrinogen | 7102912100 | 7102912500 |
Anti-HSA | 7102972100 | 7102972100 |
Anti-Transferrin | 7103062100 | 7103062500 |
Anti-HGH | 7103160100 | 7103160500 |
Anti-FSH | 7103180100 | 7103180500 |
Anti-Prothrombin | 7103452100 | 7103452500 |
Anti-EPO | 7103372100 | 7103372500 |
Anti-C1-Inhibitor | 7103402100 | 7103402500 |
Anti-Gonadotropin | 7103412100 | 7103412500 |
Recombinant protein detection and quantitation using C-tag
CaptureSelect Biotin Anti-C-tag Conjugate consists of a 13 kDa VHH affinity ligand with high affinity and selectivity for the 4-amino-acid 'C-tag' peptide tag E‐P‐E‐A (glutamic acid-proline-glutamic acid-alanine).
The Anti-C-tag affinity ligand recognizes the E-P-E-A peptide when this tag is fused directly to the C-terminus of a protein. The affinity ligand—constructed as a mono-specific bihead—is chemically conjugated to its specific label via an appropriate spacer that retains the binding reactivity of the ligand.
CaptureSelect ligand | Biotin | Biotin | HRP | Alexa Fluor 488 |
100 µg | 500 µg | 100 µg | ||
Anti-C-tag | 7103252100 | 7103252500 | 7303072100 | 7213252100 |
AAV total capsid ELISA with anti-AAV conjugated affinity ligands
CaptureSelect Anti-AAVX Conjugate consists of a 14 kDA VHH affinity ligand. The fragment specifically binds with high affinity to adeno-associated virus particles (AAV). It demonstrates good binding reactivity towards a variety of AAV serotypes, including AAV1 to AAV8, and AAVrh10. The AAVX ligand is available with different labels, such as biotin, HRP, and Alexa Fluor 647. The biotinylated and HRP-conjugated ligands can be used to develop an AAV total capsid ELISA, to quantify the total number of capsids in an AAV culture or during the production and purification process.
CaptureSelect conjugated ligands for viral vectors
CaptureSelect ligand | Biotin | Biotin | HRP | HRP | Alexa Fluor 647 |
100 µg | 500 µg | 100 µL | 500 µL | 100 µg | |
Anti-AAVX | 7103522100 | 7103522500 | 7303522100 | 7303522500 | 7233522100 |
Anti-AAV8 | 7103382100 | 7103382500 | - | - | - |
Anti-AAV9 | 7103332100 | 7103332500 | 7303332100 | 7303332500 | 7233322100 |
Anti-AdV5 | 7103532100 | 7103532500 | - | - | - |
The Human IgG-Fc PK Biotin Conjugate consists of a 13 kDa recombinant VHH affinity ligand, chemically conjugated to biotin. The ligand specifically binds to the Fc part of all four human IgG subclasses. The conjugate can be used for the detection, quantification, and characterization of all human (recombinant) IgG antibodies and Fc-fusion proteins in, for example, non-human plasma and/or serum samples like mouse, rat, rhesus, and cynomolgus monkey. This makes the conjugate very suitable for setting up pharmacokinetics (PK) assays.
CaptureSelect human kinetics biotin conjugates are bi-functional ligand constructs consisting of two different recombinant VHH affinity ligands. These conjugates enable high-affinity capture of Human IgG Fab fragments that contain a kappa or lambda light chain. The bi-functional ligands are chemically conjugated to biotin, which allows immobilization on streptavidin-functionalized surfaces. They can be used for the screening of biomolecular interactions between Fab fragments and their specific target antigen. The kinetics ligands bind the CL-kappa or CL-lambda (light chain) and the CH1 domain (heavy chain) of a human IgG Fab fragment simultaneously. This binding results in extreme low dissociation rates (Kd) between the ligand and the captured Fab fragment.
CaptureSelect conjugated ligands for kinetic experiments
CaptureSelect ligand | Biotin | Biotin |
100 µg | 500 µg | |
Anti-IgG-FC PK | 7103322100 | 7103322500 |
Anti-Human Fab-Kappa Kinetics | 7103302100 | 7103302500 |
Anti-Human Fab-Lambda Kinetics | 7103312100 | 7103312500 |
CaptureSelect conjugated ligands for exosomes
CaptureSelect Biotin and HRP exosome conjugates bind with high affinity and selectivity toward the tetraspanin markers (CD81/CD63 and CD9) expressed on extracellular vesicles (EVs) like exosomes.
Applications for these include sandwich ELISA, Gyrolab™-based immunoassays, immunoprecipitation, sample prep purification using streptavidin-coupled Dynabeads or MSIA Streptavidin D.A.R.T.’S and label-free detection platforms such as those based on surface plasmon resonance (SPR) and bio-layer interferometry (BLI).
CaptureSelect ligand | Biotin | Biotin | HRP | HRP |
100 µg | 500 µg | 100 µL | 500 µL | |
Anti-CD81 | 7103982100 | 7103982500 | 7303982100 | 7303982500 |
Anti-CD63 | 7103992100 | 7103992500 | 7303992100 | 7303992500 |
Anti-CD9 | 7104002100 | 7104002500 | 7304002100 | 7304002500 |
CaptureSelect biotinylated ligands:
Review our list of biotinylated ligands directed against antibodies and related derivatives. Find more detailed information on selectivity and use of these products in the product information sheets available for download on the specific product pages:
Binds human IgG and all Fab fragments thereof, through recognition of the CH1 domain on all four human IgG subclasses. | |
Binds human IgG, through recognition of the Fc region (CH3 domain) on all four human IgG subclasses. | |
Binds IgG through recognition of the Fc region on IgG of multiple species. View product sheet for species coverage. | |
Biotin Anti-IgG-Fc (Rabbit) | Binds to rabbit IgG (immunoglobulin G) from recombinant and plasma sources. The ligand shows no cross-binding to bovine, horse, sheep, goat, mouse, rat, and human IgG. Moderate cross-binding was observed towards swine IgG. |
Binds human IgA, IgA-dimers and secretory IgA (sIgA), through recognition of the Fc region on both human IgA subclasses. | |
Binds human, mouse and rat IgM. | |
Biotin Anti-IgE | binds to human IgE (immunoglobulin E) from recombinant and plasma sources, through recognition of the Fc region (CH4 domain) |
Binds all human lambda light chains (LC), through recognition of the constant domain (CL-lambda) on any human antibody, Fab fragment or free light chain. | |
Binds all human kappa light chains (LC), through recognition of the constant domain (CL-kappa) on any human antibody, Fab fragment or free light chain. | |
Binds all mouse - and rat kappa light chains (LC), through recognition of the constant domain (CL-kappa) on any mouse and rat antibody, Fab fragment or free light chain. | |
Biotin Anti-LC-lambda (Mouse) | Binds all mouse lambda light chains (LC), through recognition of the constant domain (CL-lambda) on any mouse antibody, Fab fragment or free light chain |
Biotin Anti-LC-lambda (Rat) | Binds all rat lambda light chains (LC), through recognition of the constant domain (CL-lambda) on any rat antibody, Fab fragment or free light chain |
Binds human IgG3 without any cross-binding to other human IgG subclasses or antibody isotypes. | |
Binds human IgG4 without any cross-binding to other human IgG subclasses or antibody isotypes. | |
High affinity capture of human IgG Fab fragments that possess a kappa light chain. Extreme low off-rates are achieved through avidity by targeting the CL-kappa domain of the Fab (light chain) and its IgG CH1 domain (heavy chain) simultaneously, thereby enabling Fab screening on target binding kinetics. | |
High affinity capture of human IgG Fab fragments that possess a lambda light chain. Extreme low off-rates are achieved through avidity by targeting the CL-lambda domain of the Fab (light chain) and its IgG CH1 domain (heavy chain) simultaneously, thereby enabling Fab screening on target binding kinetics. | |
Binds highly selective to free human kappa light chains and dimers thereof, through recognition of an epitope on the human CL-kappa domain that is masked in intact antibodies and Fab fragments, thereby providing a unique tool for specific detection and quantitation of free kappa light chains in (crude) antibody preps. | |
Binds highly selective to human IgG, through recognition of the Fc region on all four human IgG subclasses without cross-binding to mouse, rat, rhesus and cynomolgus monkey IgG, thereby making it extremely suitable for setting up pharmacokinetics (PK) assays. |
Review our list of biotinylated ligands directed against different proteins and virus particles. Find more detailed information on selectivity and use of these products in the product information sheets available for download on the specific product pages:
Binds the short C-tag sequence E-P-E-A (glutamic acid - proline - glutamic acid – alanine) when this affinity tag is fused at the C-terminus of a target protein. | |
Alexa Fluor™ 488 Anti-C-tag | Binds the short C-tag sequence E-P-E-A (glutamic acid - proline - glutamic acid – alanine) when this affinity tag is fused at the C-terminus of a target protein. |
Biotin Anti-FX | Binds to human Factor X and its activated form Factor Xa from plasma and recombinant sources |
Binds recombinant as well as plasma derived human Factor IX (FIX) | |
Binds recombinant (full length and B-domain deleted) as well as plasma derived human Factor VIII (FVIII), through recognition of the FVIII light chain. | |
Biotin anti-FVII | Binds recombinant as well as plasma derived human Factor VII (FVII) |
Biotin Anti-Gonadotropin | Binds to the alpha chain of recombinant as well as natural sourced hCG (human chorionic gonadotropin), LH (luteinizing hormone), TSH (Thyroid-stimulating hormone) and FSH (follicle-stimulating hormone) |
Biotin Anti-Human Albumin | Binds recombinant as well as natural sourced human serum albumin (HSA) including HSA-fusion proteins |
Biotin Anti-AAT | Binds recombinant as well as natural sourced human α1-Antitrypsin (AAT) |
Biotin Anti-C1-Inhibitor | Binds recombinant as well as natural sourced human C1-Inhibitor (C1-inh, C1 esterase inhibitor) |
Biotin Anti-Fibrinogen | Binds recombinant as well as natural sourced human Fibrinogen (coagulation factor I, FI) |
Biotin Anti-Transferrin | Binds recombinant as well as natural sourced human Transferrin |
Biotin Anti-α1-Acid Glycoprotein | Binds recombinant as well as natural sourced human α1-Acid Glycoprotein (AGP) or Orosomucoid (ORM) |
Biotin Anti-Haptoglobin | Binds recombinant as well as natural sourced human Haptoglobin (Hp) |
Biotin Anti-C3 | Binds recombinant as well as natural sourced human Complement Component 3 (C3) |
Biotin Anti-vWF | Binds recombinant as well as natural sourced human von Willebrand Factor (vWF) |
Biotin Anti-ApoA1 | Binds recombinant as well as natural sourced human Apolipoprotein A1 (ApoA1) |
Biotin Anti-α2-Macroglobulin | Binds recombinant as well as natural sourced human α2-Macroglobulin (A2M) |
Biotin Anti-Antithrombin III | Binds recombinant as well as natural sourced human Antithrombin III (ATIII) |
Biotin Anti-Prothrombin | Binds recombinant as well as natural sourced human Prothrombin (coagulation factor II, FII) |
Binds recombinant as well as natural sourced human Growth Hormone (hGH). | |
Binds recombinant as well as natural sourced human Granulocyte-Colony Stimulating Factor (G-CSF) | |
Binds recombinant as well as natural sourced human Follicle Stimulating Hormone (FSH), through recognition of an epitope specific for intact FSH only. | |
Biotin Anti-Insulin | Binds recombinant and natural-sourced human insulin and shows similar binding reactivity towards porcine- and bovine-sourced insulin. It also binds to human proinsulin, the precursor of insulin that still contains the C-peptide. |
Biotin Anti-EPO | Binds recombinant and natural-sourced human Erythropoietin (EPO). |
Biotin anti-AAV8 | Binds with high affinity to Adeno-associated virus particles of serotype 8 (AAV8). Note that it may cross-bind towards other AAV serotypes. |
Biotin anti-AAV9 | Binds with high affinity and selectivity to Adeno-associated virus particles of serotype 9 (AAV9) |
Biotin anti-AAVX | Binds with high affinity and selectivity to adeno-associated viral (AAV) particles of a wide variety of serotypes. |
Biotin anti-Adv5 | Binds with high affinity and selectivity to recombinant hexon capsomeres of Adenovirus serotype 5 (AdV5) particles as well as free hexon proteins thereof |
Related resources
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
Support
Events & Webinars
For Research Use or further manufacturing, not for diagnostic use or direct administration in humans or animals.