CaptureSelect™ Biotin Anti-LC-lambda (Human) Conjugate
CaptureSelect™ Biotin Anti-LC-lambda (Human) Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-LC-lambda (Human) Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-LC-lambda (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of human lambda light chains.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103082500500 μg
7103082100100 μg
Catalog number 7103082500
Price (USD)/ 500 µg
2,145.00
-
Add to cart
Quantity:
500 μg
Price (USD)/ 500 µg
2,145.00
Add to cart
CaptureSelect™ Biotin Anti-LC-lambda (Human) Conjugate
Catalog number7103082500
Price (USD)/ 500 µg
2,145.00
-
Add to cart
CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of human lambda light chains. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate allows you to:
  • Detect, quantitate, and characterize all human antibody isotypes and subclasses (for example, IgG1 to 4, IgM, IgA, IgD, and IgE), and Fab fragments thereof, that contain a lambda light chain; as well as recombinant human Fab fragments and/or Fab and Fab2 fragments obtained after papain or pepsin digestion
  • Avoid cross-binding with human antibodies that possess a kappa light chain or IgG from, for example, mouse, rat, or rabbit
  • Screen antibody-antigen interactions—biotin anti-LC-lambda (Hu) forms a stable [low k(diss)] complex with, for example, human IgG antibodies through bivalent binding
Applications for CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product LineCaptureSelect™
Quantity500 μg
TargetLambda Light Chain
TypeConjugate
Label or DyeBiotin & Other Haptens
Ligand TypeAntibody
Unit Size500 µg
Contents & Storage
Store at -5°C to -30°C.