Robust under stress
Use these robust, high-stability Thermo Scientific™ GC columns featuring popular GC stationary phases (e.g., 5% phenyl polycarborane siloxane, 5% phenyl polysilphenylene-siloxane, 5% diphenyl/95% dimethyl polysiloxane) that withstand maximum operating temperatures without risk of bleed or breakdown for the analyses of low polarity compounds in a wide variety of applications.
Common applications:
- Alcohols and phenols
- Aromatics
- Solvents and solvent impurities
- PCB congeners and aroclor mixes
- Semivolatiles
- Low polarity pesticides
- Free fatty acids
- Amines
Column | Recommended Application |
---|---|
TraceGOLD™ TG-5MS GC Columns | These columns employ the most widely used 5% phenyl MS phase in gas chromatography. |
TraceGOLD™ TG-5MS GC Columns with SafeGuard | The Safeguard feature of these columns extends column lifetime and ensures more consistent results over a longer period of time. |
TraceGOLD™ TG-5MT Metal GC Columns | These columns ensure greater stability and inertness at elevated temperatures even when working with highly active compounds. |
TraceGOLD™ TG-5SilMS GC Columns | The phenyl groups incorporated in the polysiloxane backbone offer improved thermal stability and reduce bleed and susceptibility to oxidation. |
TraceGOLD™ TG-5SILMS GC Columns with SafeGuard | The integrated SafeGuard feature of these columns offer the benefit of extended column lifetime. |
TraceGOLD™ TG-5MS AMINE GC Columns | These columns enable analysis of ppm levels of amines without column priming. |
TraceGOLD™ TG-5HT GC Columns | Ideal for high temperature GC applications, these columns offer extended operation up to 400°C. |
TRACE™ TR-5 GC Columns | Excellent starting columns for method development, these columns are capable of performing most required separations. |
TRACE™ TR-5MS GC Columns | These columns feature a popular GC-MS phase for many applications. |
TRACE™ TR-5HT GC Columns | These columns feature upper temperature limits as high as 400°C. |
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.