Our line of specialty hot plates includes external controlled hotplates and explosion-proof hotplates.
External-Controlled Hotplates | ||
---|---|---|
Enable uniform heating of large volumes and multiple vessels using Thermo Scientific External-Controlled Hotplates. These hotplates are ideal for a range of applications with acidic or basic aqueous solutions. An external controller operates outside fume hoods and corrosive environments. | ||
Part no. | RC2235Q | RC2240Q |
Plate shape | Square | Rectangular |
Plate material | Stainless-steel (porcelin coated) | Stainless-steel (porcelin coated) |
Plate size | 12 x 12 in. | 12 x 24 in. |
Voltage | 120V | 240V |
Temperature Range | 150° to 371°C | 150° to 371°C |
Explosion-Proof SAFE-T HP6 Hotplate | |
---|---|
Perform applications in Class I/Group D atmospheres with Thermo Scientific Explosion-Proof SAFE-T HP6 Hotplates. These hotplates feature overtemperature safety monitors, precise thermostatic controls and sealed aluminum housing for hazardous applications. | |
Part no. | HP11515B |
Plate shape | Square |
Plate material | Aluminum |
Plate size | 6.13 x 6.13 in. (15.6 x 15.6cm) |
Voltage | 120V |
Temperature Range | 22° to 38°C |
2200 Series Aluminum Top Hotplates | ||||
---|---|---|---|---|
Ideal for large-volume heating applications requiring precise temperature stability, including acid base digestion, sample drying, general reagent heating, heating TLC plates, and evaporations. | ||||
Part no. | HPA2235MQ | HPA2230MQ | HPA2245MQ | HPA2240MQ |
Plate shape | Square | Square | Rectangular | Rectangular |
Plate material | Aluminum | Aluminum | Aluminum | Aluminum |
Plate size | 12 x 12 in. (30.5 x 30.5cm) | 12 x 12 in. (30.5 x 30.5cm) | 12 x 24 in. (30.5 x 60.9cm) | 12 x 24 in. (30.5 x 60.9cm) |
Voltage | 120V | 240V | 120V | 240V |
Temperature Range | 38° to 371°C | 38° to 371°C | 38° to 371°C | 38° to 371°C |
Resources
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.