External-Controlled Hot Plate, 150°C to 371°C, Porcelain-coated Stainless Steel
Product Image
Thermo Scientific™

External-Controlled Hot Plate, 150°C to 371°C, Porcelain-coated Stainless Steel

Enable uniform heating of large volumes and multiple vessels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
RC2235Q120 V
RC2240Q240 V
Catalog number RC2235Q
Price (USD)/ Each
1,773.00
Online exclusive
1970.00 
Save 197.00 (10%)
-
Add to cart
Voltage:
120 V
Price (USD)/ Each
1,773.00
Online exclusive
1970.00 
Save 197.00 (10%)
Add to cart
External-Controlled Hot Plate, 150°C to 371°C, Porcelain-coated Stainless Steel
Catalog numberRC2235Q
Price (USD)/ Each
1,773.00
Online exclusive
1970.00 
Save 197.00 (10%)
-
Add to cart

Enable uniform heating of large volumes and multiple vessels using Thermo Scientific™ External-Controlled Hotplates. These hotplates are ideal for a range of applications with acidic or basic aqueous solutions. An external controller operates outside fume hoods and corrosive environment.

Key Features:
  • Isolation of controls allows operation in hazardous environments
  • Large 12 x 12″ (30.5 x 30.5 cm) and 12 x 24″ (30.5 x 60.9 cm) top plate surfaces accommodate up to 40 lb. (18.1 kg) loads
  • Power light indicates when control is operating
Temperature Control:

  • Large capacity with excellent temperature stabilityy
  • Temperature stability is ±5°C (9°F) over the operating temperature range Embedded heating elements maximize temperature uniformity and evenly transfer heat within ±10°C (18°F) at 150°C (300°F)
  • Percentage input controller delivers infinite temperature selection

Construction:
  • Porcelain-coated, stainless-steel top offers chemical and stain resistance
  • Chemical-resistant cover protects line cord
  • Stainless-steel case offers maximum resistance to corrosion

Includes:

5′ (1.5 m) 3-wire cord and plug

Specifications
Dimensions (L x W x H)12 in. x 24 in.
TypeHot Plate
Certifications/ComplianceCOA, COC
Max. Sample Load18 kg
ShapeRectangle
Voltage120 V
Top Plate MaterialPorcelain-coated Stainless Steel
Size12 x 12 in.
Temperature (Metric)150°C to 371°C
Wattage1600 w
Amperage13.3 A
Electrical Requirements120 V, 50/60 Hz
Frequency50/60 Hz
Unit SizeEach