Our lines of basic, standard, advanced, and specialty aluminum hot plates offer maximum heat transfer, strength and corrosion resistance. Aluminum surfaces provide excellent temperature uniformity with no risk of breakage.
Find the hotplate to meet your needs using our hot plate selection guide ›
Basic aluminum hot plates
Aluminum-Top Hotplates | ||
---|---|---|
Stay within the budget of your lab, clinic or classroom by using Thermo Scientific Aluminum-Top Hotplates. These durable hotplates are equipped with thermostatic temperature control to maintain uniform temperature across the aluminum plate surface. | ||
Part no. | HPA1915BQ | HPA1910MQ |
Plate shape | Square | Square |
Plate material | Aluminum | Aluminum |
Plate size | 6.25 sq. in. (15.9cm²) | 6.25 sq. in. (15.9cm²) |
Voltage | 120V | 240V |
Temperature Range | 38° to 371°C | 38° to 371°C |
Student Hotplates | ||
---|---|---|
Process delicate materials at low temperatures. The Thermo Scientific Student Hotplates offers a compact, benchtop design with variable control, for quick sample testing. | ||
Part no. | HP2305BQ | HP2310BQ |
Plate shape | Round | Round |
Plate material | Aluminum | Aluminum |
Plate size | 3.5 in. (9cm) | 3.5 in. (9cm) |
Voltage | 120V | 240V |
Max Temperature | 371°C | 371°C |
Standard aluminum hot plates
Cimarec+ Hotplate Series | ||
---|---|---|
Our Thermo Scientific Cimarec+ hotplate series offers exceptional safety and temperature performance for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations. | ||
Part no. | HP88857104 | HP88857108 |
Plate shape | Square | Square |
Plate material | Aluminum | Aluminum |
Plate size | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) |
Voltage | 100-120V | 230V |
Max Temperature | 300°C | 300°C |
Advanced aluminum hot plates
SuperNuova+ Hotplate Series | ||
---|---|---|
Our Thermo Scientific SuperNuova+ hotplate series provides reliable performance, dependable safety and simple operation. Units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision. | ||
Part no. | HP88857194 | HP88857198 |
Plate shape | Square | Square |
Plate material | Aluminum | Aluminum |
Plate size | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) |
Voltage | 100-120V | 230V |
Max Temperature | 300°C | 300°C |
Resources
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.