Extract DNA and RNA for infectious disease research applications
One nucleic acid extraction kit can be used for multiple microbiological organisms and infectious disease research applications. The Applied Biosystem MagMAX Prime Viral/Pathogen NA Isolation Kit combines the power of magnetic bead-based technology and advanced formulation to isolate viral, bacterial, fungal and parasitic nucleic acid from a variety of human biological specimens for downstream applications like qPCR and NGS.
When complemented with accessories for advanced workflows, adopting this kit can offer several benefits. Multiple infectious disease pathogens and sample types can be enabled with a single isolation kit. This kit is compatible with KingFisher Purification Instruments for automated extraction and is also compatible with downstream ThermoFisher assays for a complete infectious disease research workflow.
Looking to save time and help mitigate user errors?
See MagMAX Prime Viral/Pathogen NA Isolation Kit, Prefilled Plates*
*Prefilled plates only available in North America.
What are pathogens?
Pathogens are disease causing bacteria, virus and fungi which can infect animals and plants. Bacteria are microscopic pathogens that reproduce quickly upon entering the body. They tend to release toxins that can damage tissues and cause illness. Viruses, which are smaller than bacteria, invade a host cell then replicate, producing hundreds and thousands of new viruses to infect other host cells. Fungi are spore producing organisms that can sometimes cause diseases.
Researching pathogens using one solution for a multitude of sample types
To study pathogenic organisms, research labs can use a combination of kits and reagents along with automated instrumentation that support with the isolation of nucleic acids from the chosen sample type. The DNA and RNA can then be analyzed downstream using various analytical instruments and techniques. Learn more about nucleic acid purification for infectious disease research.
Product features and benefits
MagMAX Prime Viral/Pathogen NA Isolation Kit can help improve your infectious research through:
- Advanced formulation—Maximize nucleic acid yield for greater assay sensitivity
- Improve yield and achieve earlier amplification by up to 2.5 Ct when isolating some pathogenic targets*
- Lower viscosity of binding solution, making it easier to pipette than previous MagMAX solution
- Versatility in workflow options
- Optimized formulation and scripts for automation
- Streamlined and simple workflow
- Enable multiple applications across an array of infectious disease pathogens with one single sample preparation solution, thus reducing the need for multiple kits
- Three simple workflows
- One user guide to streamline workflow with easy-to-follow protocols to maximize laboratory efficiency
- Streamline your batches by running various sample types on a single plate via the same KingFisher script
- Sample type versatility—enable high-quality nucleic acid isolation from a variety of biological samples such as swabs in transport media (respiratory, genital, and lesion swabs), saliva, stool, urine, plasma, and serum
- Automation-ready—pair it with KingFisher Purification Instrument for rapid sample extraction and enable 96 samples to be processed in less than 60 minutes
- Sustainable design—From design to end-of-product life, incorporating more sustainable improvements to reduce environmental impacts
- Paperless documentation—User guide available digitally and easily accessible with a QR code printed on the box
- Less hazardous—All reagents are REACH**- compliant
- Greener surfactants—Biodegradable detergent
- Responsibly packaged—Compact product packaging size
- Flexible configuration available—Adaptable kit configurations, including user-filled and prefilled format***, accessories, and large stand-alone bottles
- Track your sample—Dye included to colorize proteinase K to track addition to sample plate
*Cycle Threshold (Ct) evaluation dependent on assay and workflow. We encourage all workflows and assay evaluations to be conducted by the customer.
**REACH is a regulation of the European Union, adopted to improve the protection of human health and the environment from the risks that can be posed by chemicals. Understanding REACH—ECHA (europa.eu)
***Prefilled kit only available in certain regions. Please contact your local representative to learn more.
Infectious disease research workflows
Example of a complete and simplified workflow for infectious disease research using the MagMAX Prime Viral/Pathogen NA Isolation Kit on KingFisher Purification System.
Collect samples
- Swabs in transport media
- Saliva
- Stool
- Urine
- Plasma
- Serum
Prepare 96 DW plates or use Prefilled Plates*
*Not available for advanced stool workflow
Leverage magnetic bead technology used in the new MagMAX Prime kit for automation-friendly isolation and purification of viral, bacterial, fungal and parasitic nucleic acids on KingFisher instruments.
Perform qPCR detection using QuantStudio PCR system and a variety of downstream assays for various applications
Analyze data
Collect samples
- Swabs in transport media
- Saliva
- Stool
- Urine
- Plasma
- Serum
Prepare 96 DW plates or use Prefilled Plates*
*Not available for advanced stool workflow
Leverage magnetic bead technology used in the new MagMAX Prime kit for automation-friendly isolation and purification of viral, bacterial, fungal and parasitic nucleic acids on KingFisher instruments.
Perform qPCR detection using QuantStudio PCR system and a variety of downstream assays for various applications
Analyze data
Performance data
MagMAX Prime Viral/Pathogen NA Isolation Kit provides accurate results with minimal risk to cross contamination
Basic workflow confirmed the amplification of nucleic acid across array of known bloodborne, respiratory, sexually transmitted/urinary tract pathogen targets at varying concentrations
Figure 2. *Dried Blood Spots were lysed utilizing SKU A53651 as an upfront digestion accessory to the basic workflow.
Advanced lysis and advanced stool workflows confirmed the amplification of nucleic acid across an array of known sexually transmitted/urinary tract pathogen and gastrointestinal targets at varying concentrations
Figure 3. Achieve consistent qPCR results for various molecular applications using advanced workflows by leveraging accessories like the MagMAX Prime Viral/Pathogen G+ Bacterial and Fungal Lysis Buffer for fungus and gram-positive bacteria and the MagMAX Prime Viral/Pathogen Stool Lysis Buffer with MagMAX Prime Viral/Pathogen Bead Beating Tubes for stool samples.
Prefilled kits saved the user an average of 10 mins compared to user-filled kits
Extract nucleic acids using an automated platform in ≤45 minutes with a prefilled workflow*
*Prefilled kit is only available in certain regions. Please contact your sales representative to learn more.
Ordering information
Know which additional accessory to select when working with difficult pathogen types.
Available reagents and wash solutions
See below for a list of available reagents and additional solutions for the MagMAX Prime Viral/Pathogen NA Isolation Kit
- Applied Biosystems MagMAX Prime Viral/Pathogen NA Isolation Kit
- Applied Biosystems MagMAX Prime Viral/Pathogen NA Isolation Kit, Prefilled Plate
- Applied Biosystems MagMAX Prime Viral/Pathogen Binding Beads
- Applied Biosystems MagMAX Prime Viral/Pathogen Elution Buffer
- Applied Biosystems MagMAX Prime Viral/Pathogen Wash I Solution
- Applied Biosystems MagMAX Prime Viral/Pathogen Binding Solution
- Applied Biosystems MagMAX Prime Viral/Pathogen Proteinase K
- Applied Biosystems MagMAX Prime Viral/Pathogen Proteinase K dye
- Applied Biosystems MagMAX Prime G+ Bacteria and Fungal Lysis Buffer, 1000
- Applied Biosystems MagMAX Prime Stool Lysis Buffer
- Applied Biosystems MagMAX Prime Bead Beating Tubes
- Applied Biosystems MagMAX Prime G+ Bacteria and Fungal Lysis Buffer, 100
Infectious disease testing with real-time PCRReliable solutions for respiratory, gastrointestinal, urinary tract, vaginal and sexually transmitted infections, arbovirus and more. |
For Research Use Only. Not for use in diagnostic procedures.