The MagMAX Prime Viral/Pathogen NA Isolation Kit, prefilled plates, is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, yeast, and fungal nucleic acid from biological specimens.
Features of the MagMAX Prime Viral/Pathogen NA Isolation Kit, prefilled plates, include:
• Compatible with multiple downstream assays
• Automation-ready prefilled plates that save operators time and effort when preparing for a purification run
• Improved reagent that maximizes nucleic acid yield from routinely tested pathogens without the need for additional lysis
• Streamlined workflow accommodating various sample types and scalable sample input volume that can be run on a single plate
• Single purification protocol for various biological specimens including swabs, serum, plasma, and urine
• Designed to work with KingFisher Flex and KingFisher Apex magnetic particle processors with ability to purify 96 samples in under 60 minutes
• Sustainable product with biodegradable ingredients and reduced plastic usage
• Available accessories with optimized protocol and KingFisher script to extract nucleic acids from challenging organisms and stool sample type
The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in automated magnetic bead purification techniques.
• 160 mL Binding Solution
• 300 mL Wash I Solution
• 6 mL Binding Beads
• 3 mL Proteinase K
• 36 mL Elution Buffer
• 30 μL Proteinase K dye
Store at 15–25°C.