MagMAX™ Prime Stool Lysis Buffer
MagMAX™ Prime Stool Lysis Buffer
Applied Biosystems™

MagMAX™ Prime Stool Lysis Buffer

MagMAX Prime Stool Lysis Buffer is specifically designed to enable efficient, high-throughput nucleic acid extraction from stool samples. It isRead more
Have Questions?
Catalog number A58154
Price (USD)/ 80 mL
58.65
Special Offer
Online exclusive
Ends: 16-Dec-2024
69.00 
Save 10.35 (15%)
-
Add to cart
Price (USD)/ 80 mL
58.65
Special Offer
Online exclusive
Ends: 16-Dec-2024
69.00 
Save 10.35 (15%)
Add to cart
MagMAX™ Prime Stool Lysis Buffer
Catalog numberA58154
Price (USD)/ 80 mL
58.65
Special Offer
Online exclusive
Ends: 16-Dec-2024
69.00 
Save 10.35 (15%)
-
Add to cart

MagMAX Prime Stool Lysis Buffer is specifically designed to enable efficient, high-throughput nucleic acid extraction from stool samples. It is designed to be used in conjunction with the MagMAX Prime bead-beating tubes and MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

For Research Use Only.
Specifications
Bead TypeMagnetic
ColorClear
FormatBottle
Isolation TechnologyBeads
Sample TypeFecal Swab in Transport Media, Stool
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 100 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX™
Product TypeReagent
Quantity80 mL
Sample Volume400 μL
Shipping ConditionRoom Temperature
Shelf Life2 Years
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
Unit Size80 mL
Contents & Storage
Store at 15–25°C.