Single-tube multiplex testing
Targets: 1–4
For Research Use Only. Not for use in diagnostic procedures.
Looking to expand your molecular research menu for broader pathogens? We offer a variety of testing options to suit your needs.
Targets: 1–4
Targeted Panels | |
---|---|
Examples: | |
Chlamydia trachomatis | Mycoplasma genitalium |
Neisseria gonorrhoeae | Trichomonas vaginalis |
Targets: 4–9
Targeted Panels | |
---|---|
Examples: | |
Chlamydia trachomatis | Ureaplasma parvum |
Mycoplasma genitalium | Ureaplasma urealyticum |
Mycoplasma hominis | Herpes simplex virus type 1 and 2 (HSV1 and HSV2) |
Neisseria gonorrhoeae | Trichomonas vaginalis |
For Research Use Only. Not for use in diagnostic procedures.
Real-time PCR products with high analytical sensitivity are needed to support laboratory research to study outbreaks and determine disease etiology where pathogens are similar.
Applied Biosystems TrueMark real-time qPCR solutions are part of a sample-to-results workflow. Benefits include the ability to:
Our portfolio of real-time PCR sexually transmitted infections and vaginal health research products are used to analyze a range of pathogens including viruses, bacterias, parasites and fungi, with high analytical sensitivity.
These solutions are efficient, generate informative results, and solve many of the challenges associatedwith the use of immunoassay-based, culture-based, and other molecular research methods.
Looking for ways to incorporate STI testing into your laboratory?
Applied Biosystems TrueMark STI Select Panel, Combo Kit
Format: Single-tube multiplex
Targets
TrueMark STI Select Panel | |
Bacterial | Chlamydia trachomatis Neisseria gonorrhoeae Mycoplasma genitalium |
Parasitic | Trichomonas vaginalis |
Kit size | 200 reactions |
---|---|
Throughput | 1–94 samples on 96-well plates, 1-382 samples on 384-well plates |
Control(s) | RNase P process control and Applied Biosystems TrueMark STI Amplification positive control |
Sample type(s) | Varies |
Turnaround time | ~4 hours |
Instrument compatibility |
|
Software | Applied Biosystems QuantStudio Design and Analysis Software v2.6 or later |
Cat. no. |
Applied Biosytems TrueMark STI Plus Combo Kit
Format: Pre-spotted 96-well plate
Targets
TrueMark STI Plus Panel | |
Bacterial | Chlamydia trachomatis Mycoplasma genitalium Mycoplasma hominis Neisseria gonorrhoeae Ureaplasma parvum Ureaplasma urealyticum |
Viral | Herpes simplex virus type 1 and 2 (HSV1 and HSV2) |
Parasitic | Trichomonas vaginalis |
Kit size | 60 samples/per kit |
---|---|
Throughput | 1–94 on 96-well plates |
Control(s) | RNase P process control and Applied Biosystems TrueMark Amplification positive control |
Sample type(s) | Varies |
Turnaround time | ~4 hours |
Instrument compatibility |
|
Software | Applied Biosystems QuantStudio Design and Analysis Software v2.6 or later |
Catalog number | TrueMark STI Plus Panel A56291C |
Applied Biosystems TrueMark STI and Vaginal Panels, TaqMan Array Cards
Targets
Choose up to 48 different microbial targets to create custom panels
Number of targets | 24 or 48 |
---|---|
Throughput | 8 samples/card |
Control(s) | RNase P process control and TrueMark Amplification positive control |
Sample type(s) | Varies |
Turnaround time | ~4 hours |
Instrument compatibility |
|
Software | Applied Biosystems QuantStudio Design and Analysis Software v2.6 or later |
Catalog number | Varies |
Applied Biosystems TrueMark Vaginal and STI Expanded Panel, OpenArray Plate
Targets
TrueMark Vaginal and STI Expanded Panel, OpenArray Plate | ||
Bacterial | Atopobium vaginae Bacterial vaginosis–associated bacteria 2 (BVAB2) Bacteroides fragilis Chlamydia trachomatis Enterococcus faecalis Escherichia coli O18 Gardnerella vaginalis Haemophilus ducreyi Lactobacillus crispatus Lactobacillus gasseri Lactobacillus iners Lactobacillus jensenii | Mobiluncus curtisii Mobiluncus mulieris Mycoplasma genitalium Mycoplasma hominis Neisseria gonorrhoeae Prevotella bivia Staphylococcus aureus Streptococcus agalactiae (group B) Treponema pallidum Uncultured Megasphaera 1 Uncultured Megasphaera 2 Ureaplasma urealyticum |
Viral | Herpes simplex virus type 1 (HSV1) Herpes simplex virus type 2 (HSV2) | |
Fungal | Candida albicans Candida dubliniensis Candida glabrata Candida krusei Candida lusitaniae Candida parapsilosis Candida tropicalis | |
Parasitic | Trichomonas vaginalis |
Number of targets | 34 |
---|---|
Throughput | 24 samples/plate |
Control(s) | RNase P process control and Applied Biosystems TrueMark Vaginal Microbiota Amplification positive control |
Sample type(s) | Varies |
Turnaround time | ~5 hours |
Instrument compatibility | QuantStudio 12K Flex Real-Time PCR System |
Software | QuantStudio 12K Flex Software |
Catalog number | TrueMark Vaginal and STI Expanded Panel, OpenArray Plate A39899 (Custom panel options also available) |
Expand your molecular testing research menu using the PCR instrument you have.
Learn more about PCR testing formats like single tube and plates ›
|
|
|
|
|
|
Format | Single tube assay | Duplex (target and internal control) TaqMan assays, pre-spotted | Singleplex TaqMan assays, pre-spotted | ||
Number of assays | 1 to 5 | 4 or 8 | 12 or 24 | 24 or 48 | 34 |
Number of samples | 1–384 | 12–24 | 16–32 | 1–8 | 12–48 |
Reaction volume | 10 μL, 20 μL | 10 μL, 20 μL | 10 μL | 1 μL | 33 nL |
Sample types | Varies | ||||
Instrument compatibility | • QuantStudio 5 Real-time PCR System • QuantStudio 7 Pro Real-time PCR System • Other Applied Biosystems real-time PCR instruments on request |
|
| ||
Turnaround time | <4 hours | <5 hours | |||
Software | QuantStudio Design and Analysis v2.6 or later | QS12K Flex Software | |||
Benefits | Flexibility in sample throughput; load wells only as needed | Panels with multiple targets offer an intermediate approach to testing | Fast, easy setup | Lowest cost per pathogen for broad pathogen detection studies | |
Predefined | X | X | |||
Custom |
For Research Use Only. Not for use in diagnostic procedures.
Whether you want to modify from an existing design or start from scratch, we're here to walk you through the process.
Ask about our curated, flexible content panels
Custom TrueMark Vaginal Panel (A55019)
Request a consultation or place an order › Custom TrueMark Lesion Panel (A55019)
Request a consultation or place an order › Custom TrueMark Genital Panel (A55019)
| Don't see what you're looking for? If you’re starting with your own list of targets, we offer customization across all formats to help you create a tailored solution that meets your laboratory's throughput and business needs. |
Product | Catalog number |
|
TrueMark STI and Vaginal Custom Panels | Varies | Request information |
TrueMark Antibiotic Resistance Custom Panels | Varies | Request information |
Complete your infectious disease workflow with a single kit for DNA and RNA extraction
Maximize your nucleic acid yield for greater assay sensitivity from a range of biological specimens for infectious disease applications with the MagMAX Prime Viral/Pathogen NA Isolation Kit.