Table 14.8 Molecular Probes europium and platinum luminescent FluoSpheres microspheres.
Type of Microsphere |
Size
| |
---|---|---|
0.04 µm | 0.2 µm | |
Carboxylate-Modified Microspheres | ||
Europium (365/610) | F20880 | F20881 |
Platinum (390/650) | F20886 | |
NeutrAvidin-Coated Microspheres † | ||
Europium (365/610) | F20883 | F20884 |
* Luminescent FluoSpheres microspheres are supplied as aqueous suspensions containing 0.5% solids. † See Avidin, Streptavidin, NeutrAvidin and CaptAvidin Biotin-Binding Proteins and Affinity Matrices—Section 7.6 for more information on our avidin and streptavidin conjugates. |
For Research Use Only. Not for use in diagnostic procedures.