What is a plasmid miniprep?
A plasmid miniprep is a molecular biology technique used to extract and purify small amounts of plasmid DNA from bacterial cells. Plasmids are circular DNA molecules found in bacteria, and the miniprep process aims to isolate the plasmid DNA for further analysis or use in genetic engineering experiments. The procedure involves lysing the bacterial cells, helping remove impurities, and purifying the plasmid DNA using various techniques depending on the needed purity level. The resulting purified plasmid DNA can then be quantified, characterized, and stored for future use, depending on the specific requirements of the experiment.
When to use a plasmid DNA miniprep?
A miniprep scale kit should be used when attempting to purify plasmid from an overnight bacterial culture with a volume between 1–5 mL. When choosing a plasmid miniprep kit, scientists should prioritize the quality of the resulting plasmid DNA. The quality of the plasmid DNA is crucial, as it directly affects the success and accuracy of downstream applications. A high-quality plasmid DNA should have a high purity level, free from contaminants such as genomic DNA, RNA, or proteins, which can interfere with subsequent experiments. Additionally, the plasmid DNA should be intact, without any degradation or fragmentation, as this helps ensure the integrity of the genetic material being manipulated.
The Thermo Fisher GeneJET Plasmid Miniprep Kit is a spin column-based kit that utilizes silica-based membrane technology. It yields high-quality plasmid DNA, recovering up to 20 μg per isolation procedure. With a fast and convenient procedure, this kit eliminates the need for additional extraction or precipitation steps, helping provide purified DNA that is immediately ready for use in downstream applications.
The PureLink HiPure Plasmid Miniprep Kit enables scientists to isolate high yields of pure plasmid DNA from E. coli in under two hours. This kit stands out for its fast and efficient procedure, eliminating the need for organic solvents or cesium chloride. Additionally, the kit helps ensure low endotoxin levels, further enhancing the quality of the purified plasmid DNA obtained.
Use the table below to help you choose the right solution for your plasmid DNA miniprep.
Which plasmid DNA miniprep will support your research?
Order now | Order now | Order now | Order now | |
Quick and cost-effective | Most widely accepted protocol for transfection | Rapid purification with integrated lysate clarification filter | Rapid, high-throughput transfection-grade purification | |
---|---|---|---|---|
GeneJET Plasmid Miniprep Kit | PureLink HiPure Plasmid Miniprep Kit | PureLink Pro Quick96 Plasmid Kit | PureLink 96 HQ Mini Plasmid DNA Purification Kit | |
Purity grade | Molecular | Transfection | Molecular | Transfection |
Total protocol time | < 14 min | < 120 min | 45 min | 40 min |
Yield | Up to 20 µg | Up to 30 µg | Up to 15 µg | Up to 60 µg |
High-throughput | No | No | No | Yes |
Technology | Silica membrane | Anion-exchange gravity-flow column | Silica-based spin-vacuum plates | Silica-based spin-vacuum plates |
Prep size | 50, 250 preps | 25, 100 preps | 4 x 96, 24 x 96 preps | 4 x 96 preps |
Explore: Plasmid DNA Midiprep
Explore: Plasmid DNA Maxiprep
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
- Protocol Videos—Videos to help you isolate nucleic acids using a variety of techniques.
- DNA & RNA Selection Guide—Find the right purification products.
- Videos—View our library of DNA & RNA purification and analysis videos.
- Application Notes—Explore our application notes from scientists sharing data for isolation products