Discover MagMAX Technology and the power of magnetic bead-based kits for nucleic acid purification. Purify high-quality nucleic acids from a variety sample types. Save time and increase consistency by using with KingFisher instruments for automated sample purification.
Learn about our featured MagMAX kits
The following products are recommended for research based viral nucleic acid isolation for SARS Cov-2. To learn about the differences between the kits, please visit this page.
For more information on ordering MagMAX Viral/Pathogen II Nucleic Acid Isolation kit (A48383R) please contact customer service.
MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (MVP I) Order now |
| MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit Order now | |
Who is it for? | These MagMAX Viral/Pathogen Nucleic Acid Isolation Kits are designed for use by labs that are researching SARS-CoV-2 and other infectious disease pathogens. | ||
Sample types | MVP I and MVP II: Universal viral transport media (VTM), bronchoalveolar lavage (BAL) and other respiratory research samples, serum, plasma, urine, CSF, whole blood, and saliva | MVP Ultra: Blood, Urine, Swabs, Lavage, Universal viral transport media (VTM), ThinPrep and Eswab | |
Automation level | Automation-ready protocols designed for Thermo Scientific KingFisher Apex, KingFisher Flex and KingFisher Duo Prime systems | ||
Sample input volume | 200 µL of VTM sample recommended | Sample volume inputs from 200 µL to 2 mL of recommended sample | |
Elution volume | 50 μL | Elution volumes ranging from 50 to 100 µL | |
Time per run | MVP I and MVP II: 96 samples can be processed in <40 minutes | MVP Ultra: 96 samples can be processed in <60 minutes | |
Protocol update |
| — | |
What is the difference between the two kits? | Binding Beads: In order to support the unprecedented demand for RNA extraction products for COVID-19 testing, there has been a raw material change in the bead component contained in the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit. The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit contains these beads. The chemistry driving the RNA to the beads is identical, and the nucleic acid binding capacity of each bead type is sufficient to provide similar results between the two products. Bulk reagent format: The MagMAX Viral/Pathogen II kit is only available in bulk prep format with 1,000 or 2,000 reactions. | The MVP Ultra kit includes a proprietary enzyme mix that enables lysis and nucleic acid recovery from the toughest pathogens, such as gram-positive bacteria, yeast, and fungi. | |
Cat. No. | A48310 (up to 2,000 preps/kit) This kit contains the following SKUs: A42359, A42360, A42362, A42363, A42364 | A48383R (up to 2,000 preps/kit) | A42356 (up to 100 preps) |
A42352 (up to 200 preps/kit) |
DNA | DNA | Cell-free DNA |
---|---|---|
MagMAX DNA Multi-Sample Ultra Kit 2.0 | MagMAX DNA Multi-Sample Ultra Kit | MagMAX Cell-free DNA Isolation Kit |
Faster, easier purification for saliva, blood, buffy coat
| DNA extraction for a variety of sample types
| Rapid isolation of cell-free DNA from liquid samples
|
Order Now Learn more | Order Now Learn more | Order Now Learn more |
Total RNA | mRNA |
---|---|
MagMAX mirVana Total RNA | Dynabeads mRNA DIRECT Purification Kit |
Purify true total RNA (small RNA enriched total RNA) from a variety of sample types
| Simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues |
Order Now Learn more | Order Now Learn more |
Cell-free DNA | Cell-free Total Nucleic Acid |
---|---|
MagMAX Cell-free DNA Isolation Kit | MagMAX Cell-Free Total Nucleic Acid Isolation Kit |
Rapid isolation of cell-free DNA from liquid samples
Automation ready, phenol-free extraction | Isolate and purify DNA and RNA from cell-free plasma samples
|
Order Now Learn more | Order Now Learn more |
MagMAX FFPE DNA/RNA Ultra Kit |
Sequentially extract DNA and RNA into separate eluates from the SAME FFPE section
|
Order Now Learn more |
DNA | RNA |
---|---|
MagMAX Plant DNA Isolation Kit | MagMAX Plant RNA Isolation Kit |
Automation-ready purification of DNA from a wide variety of plant species | Automation-ready purification of RNA from a wide variety of plant species |
Order Now Learn more | Order Now Learn more |
The following products are recommended for research based viral nucleic acid isolation for SARS Cov-2. To learn about the differences between the kits, please visit this page.
For more information on ordering MagMAX Viral/Pathogen II Nucleic Acid Isolation kit (A48383R) please contact customer service.
MagMAX Viral/Pathogen Nucleic Acid Isolation Kit (MVP I) Order now |
| MagMAX Viral/Pathogen Ultra Nucleic Acid Isolation Kit Order now | |
Who is it for? | These MagMAX Viral/Pathogen Nucleic Acid Isolation Kits are designed for use by labs that are researching SARS-CoV-2 and other infectious disease pathogens. | ||
Sample types | MVP I and MVP II: Universal viral transport media (VTM), bronchoalveolar lavage (BAL) and other respiratory research samples, serum, plasma, urine, CSF, whole blood, and saliva | MVP Ultra: Blood, Urine, Swabs, Lavage, Universal viral transport media (VTM), ThinPrep and Eswab | |
Automation level | Automation-ready protocols designed for Thermo Scientific KingFisher Apex, KingFisher Flex and KingFisher Duo Prime systems | ||
Sample input volume | 200 µL of VTM sample recommended | Sample volume inputs from 200 µL to 2 mL of recommended sample | |
Elution volume | 50 μL | Elution volumes ranging from 50 to 100 µL | |
Time per run | MVP I and MVP II: 96 samples can be processed in <40 minutes | MVP Ultra: 96 samples can be processed in <60 minutes | |
Protocol update |
| — | |
What is the difference between the two kits? | Binding Beads: In order to support the unprecedented demand for RNA extraction products for COVID-19 testing, there has been a raw material change in the bead component contained in the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit. The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit contains these beads. The chemistry driving the RNA to the beads is identical, and the nucleic acid binding capacity of each bead type is sufficient to provide similar results between the two products. Bulk reagent format: The MagMAX Viral/Pathogen II kit is only available in bulk prep format with 1,000 or 2,000 reactions. | The MVP Ultra kit includes a proprietary enzyme mix that enables lysis and nucleic acid recovery from the toughest pathogens, such as gram-positive bacteria, yeast, and fungi. | |
Cat. No. | A48310 (up to 2,000 preps/kit) This kit contains the following SKUs: A42359, A42360, A42362, A42363, A42364 | A48383R (up to 2,000 preps/kit) | A42356 (up to 100 preps) |
A42352 (up to 200 preps/kit) |
DNA | DNA | Cell-free DNA |
---|---|---|
MagMAX DNA Multi-Sample Ultra Kit 2.0 | MagMAX DNA Multi-Sample Ultra Kit | MagMAX Cell-free DNA Isolation Kit |
Faster, easier purification for saliva, blood, buffy coat
| DNA extraction for a variety of sample types
| Rapid isolation of cell-free DNA from liquid samples
|
Order Now Learn more | Order Now Learn more | Order Now Learn more |
Total RNA | mRNA |
---|---|
MagMAX mirVana Total RNA | Dynabeads mRNA DIRECT Purification Kit |
Purify true total RNA (small RNA enriched total RNA) from a variety of sample types
| Simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues |
Order Now Learn more | Order Now Learn more |
Cell-free DNA | Cell-free Total Nucleic Acid |
---|---|
MagMAX Cell-free DNA Isolation Kit | MagMAX Cell-Free Total Nucleic Acid Isolation Kit |
Rapid isolation of cell-free DNA from liquid samples
Automation ready, phenol-free extraction | Isolate and purify DNA and RNA from cell-free plasma samples
|
Order Now Learn more | Order Now Learn more |
MagMAX FFPE DNA/RNA Ultra Kit |
Sequentially extract DNA and RNA into separate eluates from the SAME FFPE section
|
Order Now Learn more |
DNA | RNA |
---|---|
MagMAX Plant DNA Isolation Kit | MagMAX Plant RNA Isolation Kit |
Automation-ready purification of DNA from a wide variety of plant species | Automation-ready purification of RNA from a wide variety of plant species |
Order Now Learn more | Order Now Learn more |
Discover MagMAX Technology and the power of magnetic bead-based kits for nucleic acid purification
- Superior binding capacity – the large surface area of a magnetic bead and thorough exposure to the target during mixing enables superior binding and washing efficiency
- Less clogging – due to mobile dispersion and regular density
- Improved elution – efficient elution in small volumes
- Greater sample range – efficient removal of inhibitors from a wide range of samples with different viscosities
- Versatility – suitable for low to high throughput applications
Automate MagMAX Nucleic Isolation Kits using KingFisher Instruments
- Saves time – Faster processing times when combined with the KingFisher purification systems
- Less manual – fewer manual steps before loading plates onto the KingFisher instrument
- Consistent results – Reproducible recovery of high-quality RNA and DNA with less variation in each run. Compatible with many downstream applications
- Viral DNA and RNA Isolation – recommended for viral nucleic acid isolation for SARS Cov-2 to obtain high-quality nucleic acid
Don’t work in isolation - We’re here to help
Our sample prep experts are here to help with application support, method development, or any other questions you might have for your sample prep. Please contact: techservices@lifetech.com
Resources
Support
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.