Thermo Scientific Nalgene Record keeping products help protect lab records and personnel. Hard cover notebooks feature a polyethylene coated cover that's resistant to chemical corrosion from lab chemical drips and spills. Choose from a variety of permanently bound notebooks with traditional plain paper pages or pick the Nalgene PolyPaper notebook for its extremely corrosion-resistant polymer pages for added record protection.
Single sheets of Polyolefin plastic paper are also available for creating individual documents, ideal for field data collection where inclement weather can damage paper records. Rely on chemical resistant polyolefin plastic adhesive labels and Nalgene lab markers to label your lab reagents, sample containers, and both plastic and glass labware.
Select your notebook
PolyPaper Lab Notebook | Lab Notebook with fiber paper pages | Lab Notebook with fiber paper pages | Lab Notebook with fiber paper pages | Lab Notebook with fiber paper pages | Deluxe Lab Notebook |
Extremely chemical resistant PolyPaper pages | Letter size, gridded plain paper pages | Letter size, lined plain paper pages | A4 legal size, gridded plain paper pages | A4 legal size, lined plain paper pages | Double the number and acid-free gridded plain pages |
6300-1000 | 6301-1000 | 6301-2000 | 6301-3000 | 6301-4000 | 6501-1000 |
Black | Forest green cover | Burgundy cover | Forest green cover | Burgundy cover | Burgundy cover |
PolyPaper pages | Wood pulp fiber pages | Wood pulp fiber pages | Wood pulp fiber pages | Wood pulp fiber pages | Acid-free wood pulp fiber pages |
Custom size paper: 9.25 x 11.25 in. (23 x 29cm) | Letter size paper: 8.5 x 11 in. (22 x 28cm) | Letter size paper: 8.5 x 11 in. (22 x 28cm) | A4 (legal) size paper: 8.75 x 11.25 in. (22.2 x 29cm) | A4 (legal) size paper: 8.75 x 11.25 in. (22.2 x 29cm) | Letter size paper: 8.5 x 11 in. (22 x 28cm) |
Gridded | Gridded | Lined | Gridded | Lined | Gridded |
100 pages | 96 pages | 96 pages | 96 pages | 96 pages | 184 pages |
Accepts ball point pen and permanent marking pen | Accepts all writing devices | Accepts all writing devices | Accepts all writing devices | Accepts all writing devices | Accepts all writing devices |
Related products
- Waterproof
- Resists most chemicals
- Roll or fold without cracking
- Stay flexible in cold temperatures
- Won't rip, stretch, or shrink
- UV light, rot, mildew resistant
- Standard hole punch ability
- Accepts most pens, markers, type or print
- Does not accept pencil
- Waterproof
- Resists most chemicals
- Perfect for labeling in the lab or field.
- Use with all writing devices except pencil
- Copier compatible
- Easy setup in most word processing label printing programs
- Labels most lab surface: plastic, glass, and metal
- Holds up to many lab chemicals
- Removes with alcohol
Additional helpful products
Right-to-Know Custom Labeling System
Create labels that identify a chemical and its properties using Thermo Scientific Nalgene Right-to-Know Custom Labeling System.
Technical support
For assistance choosing products appropriate for your application, please speak with a Nalgene Technical Support Representative team by phone at +1.585.586.8800 or (1.800.625.4327 US toll free), or email your request to
technicalsupport@thermofisher.com.
In Austria, France, Germany, Ireland, Switzerland, and the United Kingdom please contact technical support by phone at +800 1234 9696 (toll free) or +49.6184.90.6321, or email your request to techsupport.labproducts.eu@thermofisher.com.
Regulatory support: For regulatory documentation of product or material claims, please contact Nalgene Regulatory Support at RocRegSupport@thermofisher.com
Resources
- Break the Glass Habit Brochure
- Bottle and Carboy Selection Guide
- Plastic Properties Reference Magnet
- Plastic Labware Chemical Resistance Wall Poster