Maintenance of cells before PC-3 spheroid generation
After thawing from liquid nitrogen, cells were maintained in Nunclon Delta T25 cell culture flasks in Gibco DMEM medium supplemented with 10% Gibco FBS and 1% Pen-Strep for 2 passages before seeding for spheroid generation. ATCC protocol was followed for subculturing.
Materials required
- Nunclon Sphera 96-well plate
- Complete medium (Gibco DMEM medium supplemented with 10% FBS and 1% Pen-Strep)
- 1X PBS
- TrypLE reagent
- Geltrex Basement Membrane Matrix
- Centrifuge with a swinging bucket rotor and holder for 96-well plates
- Countess II Cell Counter or hemocytometer
- 15/50 ml centrifuge tubes
- Multi-channel pipette
- Sterile reagent reservoirs
Protocol for spheroid generation
- Once the flask was 70-80% confluent, medium from the flask was aspirated, the cells were washed once in 1X PBS and dissociated using 1–1.5 ml of TrypLE reagent.
- The TrypLE reagent was neutralized using 4 volumes of complete medium, and live-cell count and viability were captured using Countess II cell counting chamber. Cells with >90% viability were taken for spheroid generation.
- The stock of cells was diluted 1:10 to 1:20 in complete medium to make calculations for cell seeding density easier.
- Seeding cell number was calculated using the cell seeding calculator and cell suspension was diluted in ice cold DMEM medium.
- Geltrex matrix was then thawed completely on ice and added at a final concentration of 3% to the above cell suspension. (for lower cell densities reduce the concentration of Geltrex matrix)
- A volume of 200 μl of the cell suspension was added to each well of 96-well Sphera plates.
- Plates were then centrifuged at 1,500 rpm for 10 min at 4°C and placed in the incubator at 37°C and 5% CO2.
- Day 5-9, spheroids were ready, depending on seeding number.
Skip to Cell viability assay protocol or LIVE/DEAD visualization protocol
Cell Seeding Calculator
(as determined by the Countess Automated Cell Counter)
Number of cells to seed per well
(user-specified number)
... µL
Volume of cell suspension that containsthe specified number of cells
Tips
- Be very careful about the temperature conditions of Geltrex matrix. Keep it ice-cold at all times.
- While preparing the cell suspension make sure the media is cold. This cold media doesn’t harm the cells.
- Make sure after adding Geltrex matrix into the suspension it is mixed properly without forming bubbles.
- We have observed that centrifuging the plate after seeding cells facilitates cell aggregation leading to uniform spheroid formation consistently. However, this step is optional.
Morphology of PC-3 spheroids cultured in the presence of Geltrex matrix
150–5,000 PC-3 cells were seeded using Geltrex matrix for spheroid generation and brightfield images of spheroids at Day 3, Day 5, Day 7 and day 9 were captured using EVOS M7000 microscope under 4x magnification. Scale bar denotes 650 μm.
Morphology of PC-3 spheroids cultured in the absence of Geltrex matrix
150–5,000 PC-3 cells were seeded without using Geltrex matrix for spheroid generation and brightfield images of spheroids at Day 3, Day 5, Day 7 and day 9 were captured using EVOS M7000 microscope under 4x magnification. Scale bar denotes 650 μm.
Characterization of PC3 spheroids
Assessing cell viability using PrestoBlue-HS cell viability reagent
- Spheroids were generated following the spheroid generation protocol above. On Day 5, cell viability assay was performed using the PrestoBlue HS cell viability reagent.
- The PrestoBlue HS reagent was warmed up to room temperature. Then, 20 μl was added to each well containing 200 μl of medium using a multichannel pipette, and gently mixed by pipetting 2–3 times. Wells containing only fresh medium and the reagent were used as normalization control (blank).
- The plate was incubated at 37°C for 4 hours.
- Following this, fluorescence was read using the Varioskan LUX multimode plate reader using the following settings:
- Excitation: 560 nm; Emission: 590 nm
- 12 nm bandwidth
- Measurement time: 100 ms
- Bottom reading of plate
- Instrument temperature: 37°C
The data were exported to Microsoft Excel for analysis. Individual fluorescence values were normalized to Blank and mean of at least 6 replicates per cell seeding number were plotted using graphing and statistics software. The experiment was repeated 3 times.
Graph showing relative viability of PC-3 spheroids of Day 5 post–cell seeding. Error bars represent SEM.
Visualizing live and dead cells using LIVE/DEAD Viability/Cytotoxicity kit
- The populations of live and dead cells in Day 5 PC-3 spheroids were visualized by staining them using the LIVE/DEAD Viability/Cytotoxicity kit.
- Working solution was prepared by adding Calcein AM and EthD-1 at a final concentration of 1 μM and 2 μM respectively in fresh medium. (See our application note for more information on fluorescence staining of spheroids).
- NucBlue Live ReadyProbes reagent was added (2 drops per ml) to the working solution for nuclear staining of the spheroids. Spent medium of spheroids was changed 1:1 with working solution and incubated at 37°C for 3 hours.
- Following this, they were washed twice with PBS + 5% FBS (1:1 change), and finally resuspended in PBS + 0.5% FBS (1:1 change, each time centrifuging the plate at 1,500 rpm for 5 minutes at room temperature to settle the spheroids) to minimize background during fluorescence imaging.
- Fluorescence images of the spheroids were captured using the CellInsight CX7 high-content screening platform under 4X objective.
For Research Use Only. Not for use in diagnostic procedures.