Maintenance of MDA-MB-231 cells before spheroid generation
After thawing from liquid nitrogen, cells were maintained in Nunclon Delta T25 cell culture flasks in Gibco DMEM medium supplemented with 10% Gibco FBS and 1% Pen-Strep for 1 passage before seeding for spheroid generation. ATCC protocol was followed for subculturing.
Materials required
- Nunclon Sphera 96-well plate
- Complete medium (Gibco DMEM medium supplemented with 10% FBS and 1% Pen-Strep)
- 1X PBS
- TrypLE reagent
- Centrifuge with a swinging bucket rotor and holder for 96-well plates
- Countess II Cell Counter or hemocytometer
- 15/50 ml centrifuge tubes
- Multi-channel pipette
- Sterile reagent reservoirs
- Collagen I
Protocol for spheroid generation
- On the day of the experiment, medium from the flask was aspirated, the cells were washed once in 1X PBS and dissociated using 1–1.5 ml of TrypLE reagent.
- The TrypLE reagent was neutralized using 4 volumes of complete medium, and live-cell count and viability were captured using Countess II cell counting chamber. Cells with >90% viability were taken for spheroid generation.
- The stock of cells was diluted 1:10 to 1:20 in complete medium to make calculations for cell seeding density easier.
- Seeding cell number was calculated using the cell seeding calculator.
- Required number of cells were seeded in respective wells of the Nunclon Sphera plate using a multi-channel pipette. The final volume was maintained at 100 μl.
- The plate was centrifuged at 290 g for 3 minutes and placed in the incubator. This is Day 0.
- On Day 1, 100 μl of complete medium containing 6 μg/ml collagen I was added to each well, so that the final concentration of collagen in the medium is 3 μg/ml. The plate was centrifuged at 100 g for 3 minutes and placed back in the incubator.
- Spheroids were ready on Day 4 (>2,000 cells) to Day 7 (<2,000 cells) depending on the seeded cell number.
Skip to Cell viability assay protocol or LIVE/DEAD visualization protocol
Cell Seeding Calculator
(as determined by the Countess Automated Cell Counter)
Number of cells to seed per well
(user-specified number)
... µL
Volume of cell suspension that containsthe specified number of cells
Tips
- Fill the outermost wells of the plate with PBS to prevent evaporation of media during incubation.
- On Day 1, add the medium carefully through the sides of the wells.
- On Day 1, the medium should be at room temperature and not at 37 degrees. Also, keep the collagen I on ice until ready to use.
- We have observed that centrifuging the plate after seeding cells facilitates cell aggregation leading to uniform spheroid formation consistently. However, this step is optional.
Morphology of MDA-MB-231 spheroids
1,000–10,000 MDA-MB-231 cells were seeded for spheroid generation and brightfield images of spheroids at Day 2, Day 4 and Day 7 were captured using EVOS M5000 microscope under 4x magnification. Scale bar denotes 400 μm.
ECM makes a difference in MDA-MB-231 spheroid formation
1,000–10,000 MDA-MB-231 cells were seeded for spheroid generation with or without collagen according to the above-mentioned protocol. Brightfield images of cell aggregates and spheroids on Day 4 were captured using EVOS M5000 microscope under 4x magnification. Scale bar denotes 400 μm. The addition of collagen was found to dramatically improve the formation of spheroids.
Characterization of MDA-MB-231 spheroids
Assessing cell viability using PrestoBlue-HS cell viability reagent
- On Day 4 and Day 7, cell viability assay was performed using the PrestoBlue HS cell viability reagent.
- The PrestoBlue HS reagent was warmed up to room temperature. Then, 20 μl was added to each well containing 200 μl of medium using a multichannel pipette, and gently mixed by pipetting 2–3 times. Wells containing only fresh medium and the reagent were used as normalization control (blank).
- The plate was incubated at 37°C for 6 hours.
- Following this, the plate was centrifuged at 290g for 3 minutes, and fluorescence was read using the Varioskan LUX Multimode Microplate Reader using the following settings:
- Excitation: 560 nm; Emission: 590 nm
- 12 nm excitation bandwidth
- Measurement time: 100 ms
- Bottom optics reading
- “Circle” selection in the multipoint reading
- Instrument temperature: 37°C
The data was exported to Microsoft Excel for analysis. Individual fluorescence values were normalized to Blank and mean of at least 6 replicates per cell seeding number were plotted using graphing and statistics software. The experiment was repeated 3 times.
Graph showing relative viability of MDA-MB-231 spheroids of Day 4 and Day 7 post–cell seeding. Error bar represents SEM.
Visualizing live and dead cells using LIVE/DEAD Viability/Cytotoxicity kit
- The population of live and dead cells on Day 4 and Day 7 MDA-MB-231 spheroids were visualized by staining them using the LIVE/DEAD Viability/Cytotoxicity kit.
- Working solution was prepared by adding Calcein AM and EthD-1 at a final concentration of of 1 μM and 2 μM respectively in fresh medium (See our application note for more information on fluorescence staining of spheroids).
- NucBlue Live ReadyProbes Reagent was added (2 drops per ml) to the working solution for nuclear staining of the spheroids. Spent medium of spheroids was changed 1:1 with working solution and incubated at 37°C for 3 hours.
- Following this, they were washed twice with PBS + 5% FBS (1:1 change), and finally resuspended in PBS + 0.5% FBS (1:1 change), each time centrifuging the plate at 290g for 3 minutes at room temperature to settle the spheroids and minimize background during fluorescence imaging.
- The spheroids were then imaged using CellInsight CX7 high-content screening platform under 4X objective. Each image is a maximum intensity projection of 15 μm z-stacks (12-15 for 1,000–2,000 cells, 15-25 for 5,000–10,000 cells).
Note: With increasing spheroid diameter, there is reduced penetration of dyes and an increase in the dead cell population (red staining).
Representative images of Day 4 (left panel) and Day 7 (right panel) of MDA-MB-231 spheroids showing the live (green staining) and dead (red staining) cell population. Blue indicates nuclear staining. Scale bar = 300 µm.
For Research Use Only. Not for use in diagnostic procedures.