Introduction
Stimulation reagents, phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A, and monensin are useful to activate transcription factors for intracellular signaling and production of cytokines of many different immune cell types. Brefeldin A solution is required for intracellular retention of signaling proteins and cytokines. Brefeldin A is an inhibitor of intracellular protein transport. Incubation of cells in culture with brefeldin A leads to blockade of protein transport to the Golgi complex (GC) and accumulation of proteins in the endoplasmic reticulum (ER). Addition of brefeldin A during the last hours of in vitro activation of cells results in enhanced detection of intracellular cytokines. If performing an immunoassay on secreted protein (e.g., ELISA, western or multiplex protein detection), cells do not require treatment with brefeldin A.
Materials
- T25 or T75 sterile flasks with vented caps (e.g., Nunc EasYFlask Cell Culture Flasks, T25, filter, Cat. No. 156367)
- RPMI 1640 medium (e.g., BenchStable RPMI 1640 Medium, Cat. No. A4192301)
- Fetal Bovine Serum (e.g., Gibco Fetal Bovine Serum, Cat. No. 26140079)
- Cell Stimulation Cocktail: phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A, and monensin (e.g., Invitrogen Cell Stimulation Cocktail with or without protein transport inhibitors, Cat. No. 00-4975-93 or 00-4970-93)
- Brefeldin A solution (e.g., eBioscience Brefeldin A solution, Cat. No. 00-4506-51)
- Cell scraper (e.g., Nunc Cell Scrapers, Cat. No. 179693)
Procedure
|
|
|
|
|
|
|
|
|
Protocol tip:
Use of ready-made fixation buffers can help improve detection of cytokines and transcription factors in flow cytometry applications.
Supplemental protocol A: isolation of PBMC from whole blood
Materials
- Phosphate buffered saline (e.g., Gibco PBS (10X), pH 7.4, Cat. No. 70011044)
- 15 mL or 50 mL conical tube (e.g., Nunc 15 mL conical sterile centrifuge tubes, Cat. No. 339650)
- Ficoll-Paque® density separation medium
- Flow cytometry staining buffer (e.g., eBioscience Flow Cytometry Staining Buffer, Cat. No. 00-4222-26)
Procedure
|
|
|
|
|
|
|
|
Note:
As cells will be cultured, perform all steps using aseptic technique, and use buffers that do not contain azide.Protocol tip:
Significantly improve the accuracy of assessing cell health and concentration from freshly harvested peripheral blood mononuclear cells with automated counters. See detailed protocol on how to count PBMC using the Countess II FL Automated Cell Counter.
Supplemental protocol B: isolation of immune cells from lymphoid tissue
Materials
- 60 x 15 mm cell culture dish (e.g., Nunc Cell Culture Petri Dishes, Cat. No. 150340)
- Plastic 3-mL syringe or two frosted glass microscope slides
- Cell strainer (nylon mesh)
- 15 mL or 50 mL conical tube (e.g., Nunc 15 mL conical sterile centrifuge tubes, Cat. No. 339650)
- Flow cytometry staining buffer (e.g., eBioscience Flow Cytometry Staining Buffer, Cat. No. 00-4222-26)
Procedure
|
|
|
|
|
Note:
Mechanical disruption of lymphoid tissue is generally sufficient to release cells into a single-cell suspension.Note:
As cells are to be cultured, perform all steps using aseptic technique, and use buffers that do not contain azide.For Research Use Only. Not for use in diagnostic procedures.