Multiple formats for wound pathogen detection and antibiotic resistance research solutions are available to fit your laboratory needs. Broad panel testing is available on TaqMan Array Cards or OpenArray plates for low or high throughput needs, and small panel, custom solutions are available in single-tube assays and 96- or 384-well plates.
Product |
Custom SKU |
Study name |
Product format |
TrueMark Wound+ABR Expanded OpenArray Plate |
4471120 ID MA47VXD |
Wound+ABR Expanded V2 |
Format 56 No replicates
Qty 1 = 10 pk Minimum 1 |
TrueMark Wound+ABR Expanded TaqMan Array Card |
4398986 |
Wound+ABR Expanded Panel V2 |
Format 48 No replicates
Qty 1 = 1 TAC Minimum 10 |
TrueMark Wound+ABR Amplification Control |
A50377 |
|
1 tube x 100 uL 1 x 105 copies/uL
Qty 1 |
TrueMark Wound+ABR AV Control |
A50381 |
|
50 uL per tube 5 x 107 copies/uL
Qty 1 |
Product |
Custom SKU |
Study name |
Product format |
TrueMark Wound Expanded OpenArray Plate |
4471120 ID MAFVKRM |
Wound Expanded V2 |
Format 56 2x replicates
Qty 1 = 10 pk Minimum 1 |
TrueMark Wound Expanded TaqMan Array Card |
4398986 |
Wound Expanded Panel V2 |
Format 48 2x replicates
Qty 1 = 1 TAC Minimum 10 |
TrueMark Wound Amplification Control |
A50375 |
|
1 tube x 1,000 uL 1 x 105 copies/uL
Qty 1 |
TrueMark Wound AV Control |
A50379 |
|
50 uL per tube 5 x 107 copies/uL
Qty 1 |
Product |
Custom SKU |
Study name |
Product format |
TrueMark ABR Expanded OpenArray Plate |
4471120 ID MAEPR6P |
ABR Expanded Panel |
Format 56 2x replicates
Qty 1 = 10 pk Minimum 1 |
TrueMark ABR Expanded TaqMan Array Card |
4398986 |
ABR Expanded Panel |
Format 48 2x replicates
Qty 1 = 1 TAC Minimum 10 |
TrueMark ABR Amplification Control |
A50375 |
|
1 tube x 1,000 uL 1 x 105 copies/uL
Qty 1 |
TrueMark ABR AV Control |
A50379 |
|
50 uL per tube 5 x 107 copies/uL
Qty 1 |
Cat. No. |
Description |
Dye |
Scale |
A50136 |
TaqMan Antibiotic Resistance Assay |
FAM |
250 reactions |
A50137 |
TaqMan Microbe Detection Assay |
||
A52079 |
TaqMan Antibiotic Resistance Assay, pooled |
||
A52083 |
TaqMan Microbe Detection Assay, pooled |
||
Use the Microbe Assay Search Tool to find the correct Target + Assay ID combo for ordering.
|
|||
Looking for a single-tube multiplex solution? Contact your sales representative.
|
Easily select and build custom wound pathogen and ABR panels in your preferred research format, providing flexibility to meet the unique needs of your laboratory.
Choose from an extensive list of microbe assays and antibiotic resistance assays, featuring:
For Research Use Only. Not for use in diagnostic procedures.