Analysis of regulatory RNAs like microRNA (miRNA) can help decipher complex cellular processes that may impact SARS-CoV-2 pathogenesis and immune response.
Our TaqMan miRNA solutions for SARS-CoV-2 research provide the speed, sensitivity, and scalability needed to accelerate your discoveries. Explore the miRNA workflow and learn about the benefits of our solutions:
- Simple predesigned assays that contain everything you need, optimized for your target of interest
- Flexible formats to quantify hundreds of unique miRNAs in as little as five hours
- Custom assays and array formats to meet your unique requirements
Sample RNA isolation
miRNA and non-coding RNA analysis requires total RNA isolation using methods that preserve RNA integrity and expression profiles. The MagMAX mirVana Total RNA Isolation Kit is designed for isolation of total RNA, including small RNAs such as microRNAs, from a wide variety of sample types.
Assay selection for microRNA and non-coding RNA analysis
TaqMan Advanced miRNA assays provide the highest level of sensitivity and specificity when studying miRNA for validation or focused profiling and screening studies, detecting down to 60 miRNA copies into the cDNA. Our advanced chemistry utilizes a universal reverse transcription requiring only one reverse transcription reaction to study hundreds of miRNAs.
We have two methods for sensitive, precise quantification and profiling of mature miRNAs using real-time PCR: TaqMan Advanced miRNA Assays and TaqMan MicroRNA Assays.
Learn more ›
Search for TaqMan Advanced miRNA Assays ›
Search for TaqMan MicroRNA Assays ›
In addition to miRNA assay, our portfolio includes:
- TaqMan Noncoding RNA Assays—a subset of gene expression assays for quantitative real-time PCR analysis of RNA expression
Learn more › - TaqMan Pri-miRNA Assays—quantitate primary microRNA (pri-miRNA) transcripts, the most upstream RNA molecules in the microRNA biogenesis pathway, from human, mouse, or rat species
Learn more ›
Accurately quantitate hundreds of unique miRNAs in as little as five hours with the convenience of pre-spotted TaqMan Advanced miRNA Assays in OpenArray™ cards, 384-well microfluidic cards, or 96-well plates. Products include endogenous and exogenous miRNA controls for normalization of data results. We also offer array cards featuring our TaqMan MicroRNA assay chemistry, with over 740 of the most relevant microRNA assays to choose from for human or rodent species.
Learn more ›
Panel | Description |
---|---|
TaqMan OpenArray Human Advanced MicroRNA Panel, QuantStudio 12K Flex | For high throughput analysis, generate a comprehensive miRNA profile for up 43,000 data points across 48 samples in a single working day |
TaqMan Advanced miRNA Human A and B Cards | Quantitate and profile expression of 754 unique miRNAs |
TaqMan Advanced miRNA Human Serum/Plasma Card | Quantitate and determine the expression profile of 188 unique miRNAs in one serum/plasma sample. |
TaqMan Advanced miRNA Human A and B 96-well Plates, Standard or Fast | Quantitate and profile up to 752 unique miRNAs |
TaqMan Advanced miRNA Human Serum/Plasma 96-well Plates, Standard or Fast | Quantitate and profile the expression of 188 unique miRNAs in one serum/plasma sample |
We also offer customizable 384-well microfluidic cards for microRNA analysis to enable easy and affordable profiling and/or validation of hundreds of targets. TaqMan Array Cards can be customized using any predesigned assays.
Order Custom TaqMan Array Advanced miRNA cards ›
Order Custom TaqMan Array MicroRNA cards ›
Select miRNA mimics and inhibitors
For functional miRNA analysis, we offer synthetic miRNA mimics and inhibitors for thousands of microRNAs across a wide range of species.
Learn more about miRNA mimics and inhibitors ›
Learn more about miRNA functional analysis ›
Select a master mix for microRNA and noncoding RNA analysis
Select the cDNA synthesis kit and master mix based on the type of assay.
Explore solutions for every step of the workflow:
TaqMan Advanced miRNA assays | TaqMan MicroRNA assays | TaqMan Non-Coding RNA assays |
---|---|---|
TaqMan Advanced miRNA cDNA Synthesis Kit + TaqMan Fast Advanced Master Mix | TaqMan MicroRNA Reverse Transcription Kit + TaqMan Fast Advanced Master Mix | SuperScript IV Reverse Transcriptase + TaqMan Fast Advanced Master Mix |
Try our best performing TaqMan Fast Advanced Master Mix.
Run real-time PCR and analyze data
We offer real-time PCR instruments for all research needs, from simplicity on a budget to high throughput with maximum productivity.
Learn more ›
Once you order your assays, we provide the protocols and documentation specific to your assay format for download.
Learn more ›
For data analysis, we recommend the Quantstudio Design & Analysis (DA2) app on Connect, our cloud-based platform.
Sample RNA isolation
miRNA and non-coding RNA analysis requires total RNA isolation using methods that preserve RNA integrity and expression profiles. The MagMAX mirVana Total RNA Isolation Kit is designed for isolation of total RNA, including small RNAs such as microRNAs, from a wide variety of sample types.
Assay selection for microRNA and non-coding RNA analysis
TaqMan Advanced miRNA assays provide the highest level of sensitivity and specificity when studying miRNA for validation or focused profiling and screening studies, detecting down to 60 miRNA copies into the cDNA. Our advanced chemistry utilizes a universal reverse transcription requiring only one reverse transcription reaction to study hundreds of miRNAs.
We have two methods for sensitive, precise quantification and profiling of mature miRNAs using real-time PCR: TaqMan Advanced miRNA Assays and TaqMan MicroRNA Assays.
Learn more ›
Search for TaqMan Advanced miRNA Assays ›
Search for TaqMan MicroRNA Assays ›
In addition to miRNA assay, our portfolio includes:
- TaqMan Noncoding RNA Assays—a subset of gene expression assays for quantitative real-time PCR analysis of RNA expression
Learn more › - TaqMan Pri-miRNA Assays—quantitate primary microRNA (pri-miRNA) transcripts, the most upstream RNA molecules in the microRNA biogenesis pathway, from human, mouse, or rat species
Learn more ›
Accurately quantitate hundreds of unique miRNAs in as little as five hours with the convenience of pre-spotted TaqMan Advanced miRNA Assays in OpenArray™ cards, 384-well microfluidic cards, or 96-well plates. Products include endogenous and exogenous miRNA controls for normalization of data results. We also offer array cards featuring our TaqMan MicroRNA assay chemistry, with over 740 of the most relevant microRNA assays to choose from for human or rodent species.
Learn more ›
Panel | Description |
---|---|
TaqMan OpenArray Human Advanced MicroRNA Panel, QuantStudio 12K Flex | For high throughput analysis, generate a comprehensive miRNA profile for up 43,000 data points across 48 samples in a single working day |
TaqMan Advanced miRNA Human A and B Cards | Quantitate and profile expression of 754 unique miRNAs |
TaqMan Advanced miRNA Human Serum/Plasma Card | Quantitate and determine the expression profile of 188 unique miRNAs in one serum/plasma sample. |
TaqMan Advanced miRNA Human A and B 96-well Plates, Standard or Fast | Quantitate and profile up to 752 unique miRNAs |
TaqMan Advanced miRNA Human Serum/Plasma 96-well Plates, Standard or Fast | Quantitate and profile the expression of 188 unique miRNAs in one serum/plasma sample |
We also offer customizable 384-well microfluidic cards for microRNA analysis to enable easy and affordable profiling and/or validation of hundreds of targets. TaqMan Array Cards can be customized using any predesigned assays.
Order Custom TaqMan Array Advanced miRNA cards ›
Order Custom TaqMan Array MicroRNA cards ›
Select miRNA mimics and inhibitors
For functional miRNA analysis, we offer synthetic miRNA mimics and inhibitors for thousands of microRNAs across a wide range of species.
Learn more about miRNA mimics and inhibitors ›
Learn more about miRNA functional analysis ›
Select a master mix for microRNA and noncoding RNA analysis
Select the cDNA synthesis kit and master mix based on the type of assay.
Explore solutions for every step of the workflow:
TaqMan Advanced miRNA assays | TaqMan MicroRNA assays | TaqMan Non-Coding RNA assays |
---|---|---|
TaqMan Advanced miRNA cDNA Synthesis Kit + TaqMan Fast Advanced Master Mix | TaqMan MicroRNA Reverse Transcription Kit + TaqMan Fast Advanced Master Mix | SuperScript IV Reverse Transcriptase + TaqMan Fast Advanced Master Mix |
Try our best performing TaqMan Fast Advanced Master Mix.
Run real-time PCR and analyze data
We offer real-time PCR instruments for all research needs, from simplicity on a budget to high throughput with maximum productivity.
Learn more ›
Once you order your assays, we provide the protocols and documentation specific to your assay format for download.
Learn more ›
For data analysis, we recommend the Quantstudio Design & Analysis (DA2) app on Connect, our cloud-based platform.
For Research Use Only. Not for use in diagnostic procedures.