TaqMan™ Advanced miRNA Human A and B Cards
TaqMan™ Advanced miRNA Human A and B Cards
Applied Biosystems™

TaqMan™ Advanced miRNA Human A and B Cards

The Applied Biosystems TaqMan Advanced miRNA Human A and B Cards provide the convenience of pre-spotted TaqMan Advanced miRNA assaysRead more
Have Questions?
Catalog number A31805
Price (USD)/ 754 reactions
646.65
Online exclusive
704.00 
Save 57.35 (8%)
-
Add to cart
Price (USD)/ 754 reactions
646.65
Online exclusive
704.00 
Save 57.35 (8%)
Add to cart
TaqMan™ Advanced miRNA Human A and B Cards
Catalog numberA31805
Price (USD)/ 754 reactions
646.65
Online exclusive
704.00 
Save 57.35 (8%)
-
Add to cart
The Applied Biosystems TaqMan Advanced miRNA Human A and B Cards provide the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format. This product contains two cards, enabling accurate quantitation of 754 unique miRNAs in as little as five hours. Four TaqMan Advanced miRNA Assay endogenous controls are included in each card to aid in data normalization. Details about the included assays are available in the Target List file. The assays can detect and quantify the mature form of the miRNA from 1–10 ng of total RNA from tissue.

Note: The TaqMan Advanced miRNA cDNA Synthesis Kit is required for preparation of the cDNA template that is used with the TaqMan Advanced miRNA assays.

The kit enables the analysis of:
• Multiple miRNAs from a single amplified sample
• Samples that are limited in quantity, including serum, plasma, or other biological fluids

Key features include:
Universal reverse transcription (RT)—one RT step for all TaqMan Advanced miRNA assays
Sensitive—detect as few as 60 copies of input miRNA
Specific—detect only mature miRNA and distinguish between highly homologous miRNAs
Small sample input—detect and quantify mature miRNA from as little as 1–10 ng of total RNA or 2 μL of plasma or serum
Versatile—compatible with tissue and biofluids, including serum and plasma

TaqMan Advanced miRNA cards provide all the advantages of TaqMan Advanced miRNA assays in a convenient, pre-configured microfluidic card, minimizing the experimental variability and effort required to run 384 TaqMan Advanced miRNA assays in parallel. With the TaqMan Advanced miRNA cDNA Synthesis Kit, TaqMan Advanced miRNA cards enable a comprehensive expression profile consistent with miRBase v21, providing the ideal miRNA profiling solution.

TaqMan Advanced miRNA cards are compatible with the following Applied Biosystems qPCR instruments: 7900HT, Viia7, QuantStudio 7 Flex, QuantStudio 7 Pro, and QuantStudio 12k Flex.

Recommended master mix: TaqMan Fast Advanced Master Mix

TaqMan Advanced miRNA cards require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal RT chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.

These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodPrimer-probe
Format384-well microfluidic card
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast or Standard
Label or Dye2 Color
Product LineTaqMan™
Product TypeAdvanced miRNA Human A and B Card
Quantity754 Reactions
Research CategorymiRNA
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
SpeciesHuman
For Use With (Application)miRNA analysis
Volume2 μL
Unit Size754 reactions
Contents & Storage
Two 384-well arrays

The TaqMan miRNA cards are shipped at ambient temperature. Upon arrival, store at 2–8°C and keep protected from light.