Maximize valuable bench space with our compact, lightweight dry bath / block heater, available with multiple combinations of block sizes for tube size versatility.
- Digital temperature controls and display
- Small footprint
- Built-in temperature sensing probe for outstanding temperature accuracy (≤ ±0.5°C) and uniformity (≤ ±0.5°C)
- Rapid heating rate
- An operating range of ambient +2° to 100°C (ambient at 25°C)
- Multiple combination of different size of blocks allows for maximum tube size versatility during experimental procedure
- 12V low voltage power input
These products have a 2 year warranty and are CE marked. Package contains power cord with adapters (when necessary), a block lifter, and a plastic lid.
Available Compact Dry Baths
Compact Dry Bath S | Compact Dry Bath D | |
---|---|---|
Temp range | Ambient +2° to 100°C (Ambient at 25°C) | |
Temp control accuracy | at 37°C ≤ ± 0.5°C | |
Temp uniformity | at 37°C ≤ ± 0.5°C | |
Heat up time | ≤ 16 min 30° to 100°C | ≤ 20 min 30° to 100°C |
Max 1.5mL tube capacity | 24 (4 quarter blocks) | 48 (8 quarter blocks) |
Dimensions | 7.3 x 7.3 x 1.0 in. (185 x 185 x 25mm) | 7.9 x 7.9 x 1.0 in. (200 x 200 x 25mm) |
Required accessories | Full, half, or quarter blocks | |
Weight | 2.2 lbs (<1kg) | 3.3 lbs (<1.5kg) |
Voltage (50/60Hz) | 100-120V, 200-240V | 100-120V, 200-240V |
Power | 12V DC, 5A | 12V DC 7A |
Order now | 88871001 (US) 88871003 (CN, EU, UK) | 88871002 (US) 88871004 (CN, EU, UK) |