Compact Digital Dry Bath/Block Heater
Compact Digital Dry Bath/Block Heater
Thermo Scientific™

Compact Digital Dry Bath/Block Heater

Maximize bench space with our compact, lightweight dry bath with multiple combinations of block sizes for versatility.
Have Questions?
Change viewbuttonViewtableView
Model:
Compact Dry Bath D
Compact Dry Bath S
Heating Rate:
20 min. average heating rate (30° to 100°C, ambient at 25°C)
Plug Type:
CN, EU, UK
US
Catalog number 88871004
Price (USD)/ Each
602.00
Online exclusive
669.00 
Save 67.00 (10%)
-
Add to cart
Model:
Compact Dry Bath D
Heating Rate:
20 min. average heating rate (30° to 100°C, ambient at 25°C)
Plug Type:
CN, EU, UK
Price (USD)/ Each
602.00
Online exclusive
669.00 
Save 67.00 (10%)
Add to cart
Compact Digital Dry Bath/Block Heater
Catalog number88871004
Price (USD)/ Each
602.00
Online exclusive
669.00 
Save 67.00 (10%)
-
Add to cart
Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.
  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combinations of different sizes of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
  • Two sizes to meet your application volume demands
  • Convenient and easy-to-read digital display
  • Small footprint
  • Lightweight design
  • Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
  • Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
Required Accessory:
  • Unit must be ordered with block(s)
Warranty: 1-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE, cTUVus, RoHS
Controller TypeDigital
Capacity (Metric)0.2 mL
LightingLED
Plug TypeCN, EU, UK
Temperature Range (Metric)Ambient +2° to 100°C
TypeDry Bath/Block
Heating Rate20 min. average heating rate (30° to 100°C, ambient at 25°C)
IncludesDry bath, CN, EU,and UK style plugs, block lifter, plastic lid
ModelCompact Dry Bath D
No. of Blocks2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 0.5°C
Warranty1 Year, Replacement
Dimensions (L x W x H)200 x 200 x 25 mm (7.9 x 7.9 x 1 in.)
Length (English)7.9 in.
Length (Metric)200 mm
Width (English)7.9 in.
Width (Metric)200 mm
Height (English)1 in.
Height (Metric)25 mm
Weight (English)3.3 lb.
Weight (Metric)<1.5 kg
Electrical Requirements12 V, DC, 7 A
Voltage100/240 V
Unit SizeEach