PE-Conjugated Goat Anti-Human IgG is required with LABScreen Assays for the detection of IgG. This needs to be purchased separately enabling flexibility for testing procedures.
This LABScreen control serum establishes the background value for each test batch in the detection of Class I or Class II HLA antibodies. It contains a pool of negative sera and works with the coated microbeads of LABScreen products.
Adsorb Out Beads remove reactivity to latex beads from test sera with high levels of non-specific binding. Preliminary adsorption of non-specific binding material from the serum can reduce or remove the high background in a flow cytometry test. Adsorb Out consists of a special preparation of microparticles without specific antigen coating.
PreSorb is a serum clean-up reagent that consists of particles that have been shown to remove antibodies that are reactive to latex beads and certain non-HLA antigen motifs from reference human sera. Follows a familiar protocol allowing easy integration into laboratory workflow.
PE-Conjugated Anti-Human IgM PE-conjugated anti-human IgM reacts with Fc5µ portion of the human IgM heavy chain.
IgM Positive Control Beads bind to PE-Conjugated Anti Human IgM (Secondary Antibody) to generate a positive signal.
Designed to provide breakthrough performance, higher throughput, and greater flexibility to transplant diagnostics, the LABScan3D is the most advanced IVD approved Luminex system available. Dual syringe pumps and reagent reservoirs allow for higher throughput and programmable maintenance routines. This advanced instrument is compatible with all LABScreen products.
Featured software | HLA Fusion Software | HLA Fusion Research Software |
What is it? | Powerful analysis modules are specifically designed to work with One Lambda molecular typing and antibody screening products. | Used for easy analysis of molecular typing and antibody screening results. Designed to support One Lambda Research products. |
Who uses it? | HLA Laboratory | HLA Laboratory |
Supported assays |
|
LABScreen Auto Antibodies |
Integrated with HLAMatchmaker |
YES (import and export) | YES (import and export) |
Interfaces with HistoTrac |
YES (bi-directionally integrated) | YES (bi-directionally integrated) |
Not all products are CE marked or have 510(k) clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.