iConnect injectors designed for enhanced productivity
Enhance and optimize the performance of your chromatography instruments with the wide range of quality sample injectors, valves, sample loops and accessories that we offer.
Browse our complete portfolio below, or let one of our experts help you tailor a selection specific to your application and workflow needs.
Featured TRACE 1600 Series GC video
Watch the video and see the difference
Newly designed from the ground up, the Thermo Scientific TRACE 1600 and 1610 gas chromatographs are compact, versatile systems that increase productivity and lower costs in QA/QC and routine laboratories.
iConnect Injectors for TRACE 1300 and 1600 Series GC Systems
Take advantage of the high reliability, robustness, convenience, and easy maintenance our instant connect (iConnect) injector modules.
- Full range of injection modes available to cope with the most demanding sample analysis
- Outstanding retention time stability is achieved through the IEC (Integrated Electronic Control), which guarantees industry-leading 0.001 psi control over the entire working range
- Select the iConnect injectors you need and switch between them as your analytical needs demand
iConnect Split/Splitless (SSL) Injector |
|
iConnect Split/Splitless (SSL) Backflush Injector For TRACE 1600 Series GC |
|
iConnect Helium Saver Split/Splitless (SSL) Injector |
|
iConnect Programmable Temperature Vaporizing (PTV) Injector |
|
iConnect Programmable Temperature Vaporizing (PTV) Backflush Injector For TRACE 1600 Series GC |
|
iConnect Cold On-Column (COC) Injector |
|
iConnect Gas Sampling Valve (GSV) Injector |
|
Register to attend these on-demand GC webinars
Resources
Product Literature
Application Notes
Support
CMD SchemaApp code