Avoid complicated GC configurations and rapidly introduce precise amounts of gas samples directly into capillary or packed columns with the iConnect™ Gas Sampling Valve (GSV) Injector.
Avoid complicated GC configurations and rapidly introduce precise amounts of gas samples directly into capillary or packed columns. The simple operations of the unique Thermo Scientific™ iConnect™ Gas Sampling Valve (GSV) Injector include a heated 6-port diaphragm valve, and it can mount different size sampling loops to operate in split or splitless mode with the possibility of using embedded backflush capabilities. The on-board digital pneumatics enable constant or programmed gas flows or pressures. Every operation is controlled through the user interface for the greatest ease of use.
iConnect Gas Sampling Valve Injector features
Reduce downtime, simplify maintenance, and enhance GC performance
iConnect injectors and detectors redefine GC and GC-MS flexibility and usability. In two minutes, without training or tools, you can change these modules to: