Highly efficient, 5-minute cloning with TOPO
TOPO TA in vitro transcription kits utlize the pCRII-TOPO TA vector with T7 and SP6 promoters for in vitro RNA transcription and sequencing. TOPO TA technology works by directly ligating Taq-amplified PCR products in a 5-minute, benchtop reaction yielding up to 95% recombinants.
The pCRII-TOPO TA vector features:
|
Performance and value
Each TOPO TA Cloning for in vitro transcription kit contains linearized and topoisomerase I-activated pCR™II-TOPO vector, Salt Solution, dNTPs, Control Template and Primers, M13 Forward and Reverse Primers, One Shot Chemically Competent or Electrocomp™ E. coli, S.O.C. Medium, and a supercoiled control plasmid.
Using a proofreading enzyme?
Thermostable polymerases containing extensive 3´ to 5´ exonuclease activity, such as Platinum Pfx, do not leave 3´ A-overhangs. PCR products generated with Taq polymerase have a high efficiency of cloning in the TOPO TA subcloning kits because the 3´ A-overhangs are not removed. However, if you use a proofreading polymerase or wish to clone blunt-ended fragments, you can add 3´ A-overhangs by incubating with Taq at the end of your cycling program.
Alternatively, you may want to try Zero Blunt TOPO PCR Cloning Kits. These kits offer fast and efficient cloning of blunt-end PCR products generated using thermostable, proofreading polymerases. They also utilize the ccdB gene for positive selection.
订购信息
目录号 | 产品描述 | Competent cells supplied | 反应次数 | |
---|---|---|---|---|
K460001 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot TOP10 Chemically Competent E. coli | TOP10 (genotype) | 20 | 立即订购 |
K462001 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot MAX Efficiency™ DH5α T1 Phage-Resistant Chemically Competent E. coli | MAX Efficiency™ DH5α™ T1R (genotype) | 20 | 立即订购 |
K465001 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot TOP10F' Chemically Competent E. coli | TOP10F' (genotype) | 20 | 立即订购 |
K466001 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot TOP10 Electrocomp™ E. coli | Electrocompetent TOP10 (genotype) | 20 | 立即订购 |
K461020 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot Mach1™ T1 Phage-Resistant Chemically Competent E. coli | Mach1™ T1R (genotype) | 20 | 立即订购 |
K460040 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot TOP10 Chemically Competent E. coli | TOP10 (genotype) | 40 | 立即订购 |
K462040 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot MAX Efficiency™ DH5α T1 Phage-Resistant Chemically Competent E. coli | MAX Efficiency™ DH5α™ T1R (genotype) | 40 | 立即订购 |
K465040 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot TOP10F' Chemically Competent E. coli | TOP10F' (genotype) | 40 | 立即订购 |
K466040 | TOPO TA Cloning Kit for in vitro Transcription, with One Shot TOP10 Electrocomp™ E. coli | Electrocompetent TOP10 (genotype) | 40 | 立即订购 |
仅供科研使用,不可用于诊断目的。