Learn more in The Molecular Probes® Handbook
- A Diverse Selection of Organelle Probes—Section 12.1
- Probes for Mitochondria—Section 12.2
- Probes for Lysosomes, Peroxisomes and Yeast Vacuoles—Section 12.3
- Probes for the Endoplasmic Reticulum and Golgi Apparatus—Section 12.4
- Probes for the Nucleus—Section 12.5
And visit these featured pages
Table 12.1 Molecular Probes fluorescent organelle stains.
Green-Fluorescent Probes | Yellow- and Orange- Fluorescent Probes | Red-Fluorescent Probes | Blue-Fluorescent and Other Detectable Probes | ||||
Cat. No. | Probe | Cat. No. | Probe | Cat. No. | Probe | Cat. No. | Probe |
Probes for Mitochondria:Section 12.2 | |||||||
A1372 | Nonyl acridine orange | D288 | 4-Di-1-ASP (DASPMI) | M7512 | MitoTracker Red CMXRos * | L6868 | Lucigenin † |
D273 | DiOC6(3) ‡ | D426 | DASPEI | T3168 | JC-1 § | ||
D378 | DiOC7(3) ‡ | M7510 | MitoTracker Orange CMTMRos * | D22421 | JC-9 § | ||
M7514 | MitoTracker Green FM * | R634 | Rhodamine 6G | M22425 | MitoTracker Red FM * | ||
R302 R22420 | Rhodamine 123 | T668 | Tetramethyl- rhodamine, methyl ester (TMRM) | M22426 | MitoTracker Deep Red FM * | ||
S7529 | SYTO 18 yeast mitochondrial stain | T669 | Tetramethyl- rhodamine, ethyl ester (TMRE) | ||||
T3168 | JC-1 § | ||||||
D22421 | JC-9 § | ||||||
Oxidation-Sensitive Probes for Mitochondria:Section 12.2 | |||||||
D632 | Dihydrorhodamine 123 | M7511 | MitoTracker Orange CM-H2TMRos * | M7513 | MitoTracker Red CM-H2XRos * | ||
Probes for Acidic Organelles Including Lysosomes:Section 12.3 | |||||||
L7526 | LysoTracker Green DND-26 | L12491 | LysoTracker Yellow HCK-123 | L7528 | LysoTracker Red DND-99 | H22845 | Hydroxy- stilbamidine |
L12492 | LysoTracker Deep Red | L7525 | LysoTracker Blue DND-22 | ||||
R14060 | RedoxSensor Red CC-1 ** | ||||||
pH-Sensitive Probes for Acidic Organelles:Section 12.3 | |||||||
L7535 | LysoSensor Green DND-189 | A1301 | Acridine orange | N3246 | Neutral red | L7533 | LysoSensor Blue DND-167 |
L7545 | LysoSensor Yellow/Blue DND-160 § | L7545 | LysoSensor Yellow/Blue DND-160 § | ||||
L22460 | LysoSensor Yellow/Blue 10,000 MW dextran § | L22460 | LysoSensor Yellow/Blue 10,000 MW dextran § | ||||
Probes for the Endoplasmic Reticulum:Section 12.4 | |||||||
E34250 | ER-Tracker Green | D282 | DiIC18(3) | E34251 | ER-Tracker Red | E12353 | ER-Tracker Blue-White DPX |
D273 | DiOC6(3) | D384 | DiIC16(3) | ||||
R648MP | Rhodamine B, hexyl ester | ||||||
Probes for the Golgi Apparatus:Section 12.4 | |||||||
D3521 B22650 | BODIPY FL C5-ceramide § | D3521 B22650 | BODIPY FL C5-ceramide § | ||||
N1154 N22651 | NBD C6-ceramide | D7540 B34400 | BODIPY TR ceramide | ||||
Probes for the Nucleus: Section 12.5 includes a complete listing of SYTOX, HCS NuclearMask, Hoechst, DAPI and other nuclear stains. | |||||||
* Aldehyde-fixable probe. † Chemiluminescent probe. ‡ Selective for mitochondria only at low applied concentrations (<100 nM). § Dual-emission spectrum. ** The differential distribution of the oxidized product between mitochondria and lysosomes appears to depend on the oxidation–reduction (redox) potential of the cytosol. |
For Research Use Only. Not for use in diagnostic procedures.