High sensitivity capillary HPLC for biomolecules
Use these capillary reversed phase (RP) columns for high-sensitivity HPLC-UV or LC/MS characterization of intact proteins in top-down proteomics applications where sample amount is critical.
- Intact proteins in top-down proteomics applications
- Monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs)
- Bio-specific proteins
Thermo Scientific capillary HPLC Columns for biomolecules
Thermo Scientific capillary HPLC columns have excellent resolution, long column lifetime, and low carry-over.
MAbPac RP Capillary HPLC Column
Take advantage of the standalone for low-flow HPLC and to obtain high quality data that’s comparable to high flow experiments. We recommend using the MAbPac reversed phase capillary HPLC column for:
- High sensitivity intact protein characterization
- Peptide mapping studies
- Monoclonal antibody (mAb) characterization
EASY-Spray MAbPac Capillary Column
Ensure robust nano and capillary flow LC-MS with Thermo Scientific EASY-Spray HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance.
Related products
EASY-Spray products
Nano & capillary emitters
Chromatography consumables on-demand webinar series
On-demand Consumables for chromatography webinar series: Learn at your own pace on a wide range of topics from column troubleshooting, method transfer, to tips for successful low-flow and more
Featured resources
Roland Bruderer, Principal Scientist, R&D, at Biognosys AG speaks about MAbPac RP 4 μm Stand Alone and EASY-Spray LC columns and nano and capillary flow emitters.