Nanoflow HPLC separations and nano-electrospray ionization can maximize LC-MS performance, but they rely on numerous critical and error-prone connections to achieve optimum results. Thermo Scientific™ EASY-Spray™ Source uses integrated, temperature-controlled column-emitter units that require only a single easy connection. The result is state-of-the-art performance — narrow peaks, maximum separation efficiency and outstanding MS sensitivity — for both expert and nonexpert users.
Exceptional Performance and Reliability
Broad MS Compatibility
The EASY-Spray Ion Source is available in two versions. The original EASY-Spray Ion Source works in conjunction with these Thermo Scientific mass spectrometers:
The EASY-Spray NG ion source works in conjunction with the latest next-generation Thermo Scientific mass spectrometers:
Specifications, terms and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.