Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your analyses needs.
View the relevant questions below:
Having problems with your experiment? Visit
No. When upgrading to a newer version of BioPharma Finder, DO NOT uninstall the older version before installing the current version. During the installation of the new version, the software will automatically detect the older version and make a backup copy of your old results. Then, it will uninstall the old software, install the newer version, and upgrade all of your information.The backup can be deleted or removed from your computer once you have confirmed the backup was successful.
All of the results, sequences and processing methods are saved onto the PC which the software is installed into a Mogo database. The database is automatically installed when the software is installed.
Peptide Mapping and Top Down results are saved automatically while manual intact deconvolution results must be saved by the user. However, if you process intact deconvolution experiments using the queue, the results will be saved automatically.
Need more information? Contact us ›
For Research Use Only. Not for use in diagnostic procedures.