Hot plate stirrers are popular lab tools, offered in basic, standard and advanced models. Basic models have good quality, reliability and performance, standard models come with digital display and control, as well as accurate temperature and speed control with safety protection and Advanced models feature additional timer and programmable functionality. Our hot plate stirrers are for use with all applications in the general lab.
Find the hot plate stirrer to meet your needs using our hot plate stirrer selection guide ›
Which type of hot plate stirrer fits your application best?
Basic | Standard | Advanced | |
---|---|---|---|
User Interface | Analog | Digital | Digital |
Timer | ✓ | ||
Save Settings | ✓ | ||
Hot Top Warning Light (Safety Feature) | ✓ | ✓ | |
Quick Heat-up Time to Max Temp | ✓ | ||
Accuracy of Temp and RPM Setting | Within +/-1 degree | Within +/-1 degree | |
Slow Speed Capability | ✓ | ✓ | |
Removable Plug | Round Top Only | ✓ | ✓ |
Over Temp Protection | ✓ | ✓ | ✓ |
Temperature Probe Option | ✓ | ✓ |
Single Point Hot Plate Stirrers
Single point hotplate stirrers are available in basic, standard and advanced models, offering various top plate materials and multiple sizes for all applications in the general lab.
Multi Point Hot Plate Stirrers
Multi point hot plate stirrers are ideal for high-volume, high-performance processing and life science/biotechnology applications as well as labs with multiple users.
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.