Supporting the path to detection and recovery
Explore the new expanded test menu!
- Simple and easy to use urine drug screening dipcard and cups
- Simple to Use – Easy to Read
- Test results available quickly
- Specimen validity Test results available in 2 minutes
Thermo Fisher Scientific, a market leader in drugs of abuse screening products, has partnered with Confirm Biosciences to provide simple urine drug screening solutions for the clinical market, in addition to drug courts and forensic laboratories in the United States. Detect up to 16 drugs in 5 minutes with easy-to-use urine drug screen cups!
View the full product specifications in the CLIA-waived Urine Drug Screening Dipcard and Cups Flyer
Cups Test Panel Configurations
Dip Card Test Panel Configurations
View the full product specifications in the CJ&F Urine Drug Screening Dipcard and Cups Flyer
Cups Test Panel Configurations
*Collection Kit includes: x1 Sleeves X1 Specimen Bag x1 Soap Wipe per cup
**(CJ&F) For Criminal Justice and Forensic use only in US
Dip Card Test Panel
**(CJ&F) For Criminal Justice and Forensic use only in US
End-to-end chain of custody
The Paracelsus Software from ACT Innovations drives the entire testing process. This begins with automated scheduling of participants, randomization of tests and full chain of custody along the entire workflow from sample collection to reporting of the results within Drug Court Case Management (DCCM) systems.
For technical questions and product issues regarding the urine drug screen cups, please contact Confirm Biosciences at cs@confirmbiosciences.com or 1-800-908-5603.
How to read and interpret the results: Urine drug screen cups provide a qualitative screening positive or negative result. A more specific alternative method such as LC-MS/MS or GC-MS must be used for a confirmed analytical result.
Ensuring validity of the test: There should always be a line corresponding to the control line (C). If this is not there, the test is invalid.
Positive result: No line corresponding to the test strip (T) indicates a positive result.
Negative result: Faint line or a clear line corresponding to the test strip (T) indicates a negative result.
Limitations of the test:
Where indicated, Drug Tests (Strip/Card/Device/Cup) are for professional in vitro diagnostic use, and should be only used for the qualitative detection of drugs of abuse. This assay provides a preliminary analytical test result only. A more specific alternative chemical method such as LC-MS/MS or GC/MS must be used in order to obtain a confirmed analytical result. Clinical consideration and professional judgment should be applied to any test result, particularly when preliminary positive results are indicated. There is a possibility that technical or procedural errors as well as other substances and factors may interfere with the test and cause false results.