Meet the demands of daily pipetting with the right pipette for your workflow. When you are looking for improved accuracy and precision, we have a selection of manual, electronic, single channel and multichannel pipettes.
Description | ||
The F1-ClipTip pipetting system provides a complete seal on every channel for precise sample volumes, saving valuable time to accelerate your discoveries. No longer bang tips onto your pipette. With a light touch each tip is clipped securely on the pipette and remains sealed until you decide to eject the tips. F1-ClipTip supports consistent and reproducible pipetting. | Enjoy the enhanced volume adjustment on Thermo Scientific Finnpipette F1 system pipette, which is silent and effortless with a textured adjustment knob for a firmer feel when setting volumes. The adjustable finger rest and lightweight design maximize comfort without sacrificing performance. Get optimal precision and ergonomics, all in a lightweight design that maximizes user comfort and reduces strain. | Finnpipette F2 pipettes meet the challenges of high demanding applications where robustness and durability are essential. With a sleek and rugged design the variable volume pipettes are built with tough PVDF components that stand up to harsh chemicals and damaging effects of UV light. These variable volume pipettes are fully autoclavable without disassembling. They feature a wide selection of expressly designed tips for optimal performance, precision and accuracy. |
Channels | ||
Single, 8 - , 12 -channel |
Single, 8-, 12- 16-channel |
Single, 8-, 12- 16-channel |
Format | ||
Variable and Fixed Volume | Variable and Fixed Volume | Variable and Fixed Volume |
Volume Range | ||
0.1-1000 µl (variable) 10-1000 µl (fixed) |
0.2-10,000 µl (variable) 10-10,000 µl (fixed) |
0.2-10,000 µl (variable) 10-10,000 µl (fixed) |
|
||
---|---|---|
Description | ||
E1-ClipTip Pipettes are designed for accurate and efficient liquid handling. They feature an electronic control system that supports precise pipetting volume with a simple push of a button. The ClipTip technology secures leak-free sample pickup and ejection. With it's ergonomic design and lightweight construction, the pipette offers comfortable and fatigue-free pipetting. | The Equalizer Pipette is a state-of-the-art laboratory instrument that revolutionizes liquid handling. Designed to transfer multiple samples between different labware formats using adjustable tip spacing, the Equalizer helps save time and repetitions compared to the single channel manual pipette. The Cliptip technology supports secure and leak-free sample pick up and ejection, minimizing risk of cross-contamination. | The Finnpipette Novus Pipette is a reliable and versatile laboratory tool designed for accurate and efficient pipetting. It features a lightweight and ergonomic design that supports comfortable and fatigue-free pipetting, even during extended use. With its user-friendly interface and intuitive operation, the pipette simplifies pipetting tasks and enhances productivity in the laboratory. Its durable construction and high-quality materials make it a durable and long-lasting pipette choice for scientists and researchers. |
Channels | ||
Single, 8-, 12-, 16- channel |
6-, 8-, 12- channel |
8-, 12-, 16- channel |
Volume Range | ||
0.5 – 1250 µl | 0.5 – 1250 µl | 1 – 5000 µl |
Fine Tip | Special purpose | Exra Long | Exact Volume | Capillary | |||
---|---|---|---|---|---|---|---|
Options | |||||||
Bulb draw (mL): 0.3, 0.9, 1.8, 2.2, 2.4, 2.8, 3.0, 4.7, 6.2, 7.4 |
Bulb draw (mL): 0.8, 1.8, 2.1, 3.2, 3.5, 4.3, 7.4 |
Stem diameter (mm): 2.5, 3.0, 8.0 |
Stem diameter (mm): 1.2, 3.0, 4.6, 6.1 |
Bulb draw (mL): 0.2, 3.3 |
Length: 9 in (22.5 cm) 12 in (30.0 cm) |
Dispense Volume (µl): 20, 40, 50, 60, 80, 100, 150, 200, 250, 300, 400 |
Volume (µl): 25, 50 |
Applications | |||||||
Rapid transfer, controlled drop size, calibrated transfer |
Remove separate liquids, dispense small drops |
Genomic and proteomic, reagent prep, drop additions to culture plates |
Sample preparation, layering in columns, transfers in columns, loading gels |
Assay development, research lab |
Assay development, research lab |
Assay development, research in microbiology, agriculture, veterinary, food |
Liquid transfer, rapid tests, lateral flow assay development |
Recommended Use | |||||||
Larger aperture for creams and gels, oils, industrial slurries, paints, perfumes |
Ideal for measure transfers, drop by drop work, aliquoting |
Loading gels, PCR work, DNA sample removal, feed hybridomas, lipid extraction, atomic absorption, spectroscopy |
Loading gels, PCR, lipid extractions, DNA samples removal, protein assays |
Grear for viscous fluid, paddle for mixing |
Sub-culturing and harvesting cells and tissues; Transferring from large bottle; Removal of supernatant fluids from centrifugation |
Exact volume dispensing, analytical testing Pathogen testing |
Liquid transfer, point of care tests, rapid tests, lateral flow assay development |
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.