The Thermo Scientific Talos F200X STEM is a scanning transmission electron microscope that combines outstanding high-resolution STEM and TEM imaging with industry-leading energy dispersive X-ray spectroscopy (EDS) signal detection. 2D/3D chemical characterization with compositional mapping is performed by 4 in-column SDD Super-X detectors with unique cleanliness. The Talos F200X scanning transmission electron microscope allows for the fastest and most precise EDS analysis in all dimensions, along with high-resolution TEM and STEM (HRTEM and HRSTEM) imaging with fast navigation for dynamic microscopy. The Talos F200X scanning transmission electron microscope also features reduced environmental sensitivity; the instrument enclosure moderates the impact of air pressure waves, air flows, and fine temperature variations in the TEM room.
STEM imaging and chemical analysis
The Talos F200X (S)TEM delivers fast, precise, quantitative characterization of nanomaterials in multiple dimensions. With innovative features designed to increase throughput, precision, and ease of use, the Talos F200X (S)TEM is ideal for advanced research and analysis in academic, government, semiconductor and industrial environments.
The need for large area correlative imaging at high resolution has recently increased as it allows researchers to preserve the context of their observations while also providing statistically robust data. Thermo Scientific Maps Software (enabled by Thermo Scientific Velox Software) automatically acquires an array of images across a sample and stitches them together to create one large final image. Image acquisition can even be performed unattended. The APW (Automated Particle Workflow) pack has all the benefits described in this section and adds unique processing on a dedicated processing PC with Thermo Scientific Avizo2D Software. You can get nanoparticle parameters like size, area, perimeter, shape, factor, contacts, etc., in an automated way. The fully automated and unattended software pack enables you to use the Talos F200X (S)TEM 24/7, get much better statistics and significantly improve the repeatability because operator bias is not present.
Align Genie Automation Software eases the learning curve for novice operators, reduces tensions in a multi-user environment, and improves time-to-data for the experienced operator.