Complete solution for STIs with multiplex kits
Our comprehensive portfolio supports sexual health testing with CE-marked assays for sexually transmitted infections (STI) and bloodborne infections, designed to provide accurate, reliable results with excellent sensitivity and specificity across a wide variety of bacteria such as Chlamydia trachomatis (CT), Neisseria gonorrhoeae (NG) and mycoplasma genitalium, parasites such as Trichomonas vaginalis, or viruses such as human papillomavirus (HPV) or Herpes simplex virus (HSV-1/2), Hepatitis and HIV.
Rely on our broad range of reliable and simple PCR-based testing to meet your reproductive health lab’s clinical testing needs.
Complete solution for STIs with multiplex kits
Simple laboratory workflow
Easily combinable with other PCR kits from the TaqPath Menu | GeneProof STI portfolio in one workflow
High sensitivity and specificity
Compatible with a wide range of real-time PCR devices
Contamination reduction
Ready-to-use master mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing possible carryover contamination
Easy-to-use concept
Single-tube ready-to-use master mix contains all components for PCR amplification
No additional pipetting of PCR reagents is necessary
For In Vitro Diagnostic Use.