Enzyme-linked immunosorbent assays (ELISAs) are plate-based assays for detecting and quantifying a specific protein in a complex mixture. The detection and quantification of target-specific protein in a sandwich ELISA is accomplished by using highly specific antibodies that immobilizes the target protein (antigen) to the plate and indirectly detects the presence of the target protein. This type of capture assay is called a sandwich assay because the analyte being measured is bound between two primary antibodies, each detecting a different epitope of the antigen—the capture antibody and the detection antibody. There are many different types of components, such as substrates, plates, and other reagents, that the choices can sometimes be overwhelming. We’ve made general suggestions in the protocols below, and you can also visit our ELISA builder online selection guide that asks a few simple questions about your specific ELISA and then recommends all the components you’ll need, from plates to stop solution.
Search available antibody pairs and ELISA kits Browse available ELISA reagents
Find protocols below for a standard sandwich ELISA using a 96-well plate for the detection techniques—colorimetric (chromogenic), chemiluminescent, and fluorescent detection.
Overall procedure
- Attachment of capture antibody specific to target protein to a microplate
- Addition of standards and samples containing unknown amount of the target protein which binds to the capture antibody
- Washing to remove unbound substances
- Addition of a detection antibody that binds to the immobilized target protein
- Washing away excess detection antibody and addition of HRP conjugate
- Addition of HRP substrate for indirect detection of bound protein
Colorimetric sandwich ELISA protocol
This protocol represents an example colorimetric sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP.
Materials
- Clear 96-well plate (Pierce 8-Well Polystyrene Strip Plates, Corner Notch, Cat. No. 15031)
- Coating buffer (50 mM carbonate buffer, pH 9.4, or 10 mM phosphate buffer, pH 7.4 (BupH Carbonate-Bicarbonate Buffer Packs, Cat. No. 28382)
- Blocking buffer (Assay Buffer) (SuperBlock (TBS) Blocking Buffer, Cat. No. 37535)
- Wash buffer (Tris-buffered or phosphate-buffered saline with 0.05% Tween 20 detergent) (Pierce 20X TBS Tween 20 Buffer, Cat. No. 28352, or Pierce 20X PBS Tween 20 Buffer, Cat. No. 28360)
- Plate sealers (Sealing Tape for 96-Well Plates, Cat. No. 15036)
- Reagent reservoirs (ELISA Reagent Reservoirs, Cat. No. 15075)
- Streptavidin-HRP (Pierce High Sensitivity Streptavidin-HRP, Cat. No. 21130)
- TMB substrate solution (1-Step Turbo TMB-ELISA Substrate Solution, Cat. No. 34022)
- Stop solution (0.16 M sulfuric acid) (Stop Solution for TMB Substrates, Cat. No. N600)
Additional materials required
- Capture (coating) antibody
- Biotinylated detection antibody
- Absorbance-based microplate reader (e.g., Multiskan FC Microplate Photometer)
- Distilled or deionized water
- Samples (see ELISA sample preparation protocols) and standards
Protocol tips
For a complete set of ELISA Buffers, Invitrogen Antibody Pair Buffer Kit, Cat. No. CNB0011, includes: Coating Buffer (pH 7.4 and pH 9.4), Assay Buffer (Blocking Buffer), Wash Buffer, Stabilized TMB, and Stop Solution.Protocol tips
To help ensure reliable results with accuracy and precision, high-quality washing is essential. Wash plates securely with automatic plate washers such as the Thermo Scientific Wellwash Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications.Procedure
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
Using an alkaline phosphatase system
If alkaline phosphatase (AP) is to be used instead of HRP for the enzyme conjugate, an AP-specific substrate must be used. Substitute TMB substrate solution in step 15 with p-nitrophenyl phosphate (PNPP) (Thermo Scientific 1-Step PNPP Substrate Solution, Cat. No. 37621). Incubate at room temperature for 15–30 minutes. Substitute the stop solution with 50 µL 2 N NaOH to stop the reaction. Measure absorbance of each well at 405 nm.
Performing direct antigen immobilization
When immobilizing the antigen-containing sample directly to the plate, there is no need for a capture antibody. Different concentrations of the sample should be prepared in coating buffer and identical volumes added directly to the plate. The rest of the protocol should be performed as previously described using a detection antibody and enzyme conjugate plus substrate.
Using an enzyme-labeled secondary antibody
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
Chemiluminescent sandwich ELISA protocol
This protocol represents an example of a chemiluminescent sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP. A luminol-based substrate is used for detection.
Materials
- Opaque 96-well plate (Pierce 96-well polystyrene plates, white opaque, Cat. No. 15042)
- Coating buffer (10 mM phosphate buffer, pH 7.4 or 50 mM carbonate buffer, pH 9.4) (BupH Carbonate-Bicarbonate Buffer Packs, Cat. No. 28382)
- Blocking buffer (StartingBlock T20 PBS Blocking Buffer, Cat. No. 37539, or StartingBlock T20 TBS Blocking buffer, Cat. No. 37543)
- Wash buffer (Tris-buffered or phosphate-buffered saline with 0.05% Tween 20 detergent) (Pierce 20X TBS Tween 20 Buffer, Cat. No. 28360, or Pierce 20X PBS Tween 20 Buffer, Cat. No. 28352)
- Chemiluminescent substrate (SuperSignal ELISA Pico Chemiluminescent Substrate, Cat. No. 37070)
- Streptavidin-HRP (if detection antibody is biotinylated) (HRP-Conjugated Streptavidin, Cat. No. N100)
- Reagent reservoirs (ELISA Reagent Reservoirs, Cat. No. 15075)
- Plate sealers (Sealing Tape for 96-Well Plates, Cat. No. 15036)
Additional materials required
- Capture (coating) antibody
- Detection antibody
- Luminometer-based microplate reader (e.g., Varioskan ALF Multimode Microplate Reader)
- Distilled or deionized water
- Samples (see ELISA sample preparation protocols)
Protocol tips
To help ensure reliable results with accuracy and precision, high-quality washing is essential. Wash plates securely with automatic plate washers such as the Thermo Scientific Wellwash Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications.Protocol tips
Chemiluminescence detection is recommended when detecting and quantifying low abundant proteins or when sample and primary (capture and detection) antibodies are limited.Protocol tips
White or black plates can be used for chemiluminescent detection. White plates typically display higher signal than black plates, and black plates should be used when background signal is an issue.Procedure
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
Using an alkaline phosphatase system
If alkaline phosphatase (AP) is to be used instead of HRP for the enzyme conjugate, an AP-specific substrate must be used. Substitute TMB substrate solution in step 15 with p-nitrophenyl phosphate (PNPP) (Thermo Scientific 1-Step PNPP Substrate Solution, Cat. No. 37621). Incubate at room temperature for 15–30 minutes. Substitute the stop solution with 50 µL 2 N NaOH to stop the reaction. Measure absorbance of each well at 405 nm.
Performing direct antigen immobilization
When immobilizing the antigen-containing sample directly to the plate, there is no need for a capture antibody. Different concentrations of the sample should be prepared in coating buffer and identical volumes added directly to the plate. The rest of the protocol should be performed as previously described using a detection antibody and enzyme conjugate plus substrate.
Using an enzyme-labeled secondary antibody
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
Fluorescent sandwich ELISA protocol
This protocol represents an example of a fluorescent sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP. A fluorogenic peroxidase substrate is used for detection.
Materials
- Black 96-well plate (Black 96-Well Immuno Plates, Cat. No. 437111)
- Coating buffer (10 mM phosphate buffer, pH 7.4 or 50 mM carbonate buffer, pH 9.4) (BupH Carbonate-Bicarbonate Buffer Packs, Cat. No. 28382)
- Blocking buffer (StartingBlock T20 PBS Blocking Buffer, Cat. No. 37539 or StartingBlock T20 TBS Blocking buffer, Cat. No. 37543)
- Wash buffer (Tris-buffered or phosphate-buffered saline with 0.05% Tween 20 detergent) (Pierce 20X TBS Tween 20 Buffer, Cat. No. 28352 or Pierce 20X PBS Tween 20 Buffer, Cat. No. 28360)
- Fluorogenic peroxidase substrate (QuantaBlu Fluorogenic Peroxidase Substrate Kit, Cat. No. 15169)
- Streptavidin-HRP (if detection antibody is biotinylated) (HRP-Conjugated Streptavidin, Cat. No. N100)
- Reagent reservoirs (ELISA Reagent Reservoirs, Cat. No. 15075)
- Plate sealers (Sealing Tape for 96-Well Plates, Cat. No. 15036)
Additional materials required
- Capture (coating) antibody
- Detection antibody
- Microplate fluorometer (e.g., Varioskan ALF Multimode Microplate Reader)
- Distilled or deionized water
- Samples (see ELISA sample preparation protocols)
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
Using a fluorophore-conjugated detection antibody
With a fluorophore-conjugated detection antibody, no enzyme conjugate or substrate is required. The plate fluorescence units can be measured directly after step 11. The working concentration of labeled antibody or protein is typically 2–4 μg/mL.
Performing direct antigen immobilization
When immobilizing the antigen-containing sample directly to the plate, there is no need for a capture antibody. Different concentrations of the sample should be prepared in coating buffer and identical volumes added directly to the plate. The rest of the protocol should be performed as previously described using a detection antibody and enzyme conjugate plus substrate.
Using an enzyme-labeled secondary antibody
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
Antibody dilution recommendations
The following tables provide recommended ranges for different ELISA components. Concentrations are guidelines only; for best results optimize each component individually.
Recommended starting concentration ranges for coating and detection antibodies for ELISA optimization. The use of non-purified antibodies will work but may result in higher background. It is generally recommended to use affinity-purified antibodies for optimal signal-to-noise ratio.
Source | Coating antibody | Detection antibody |
Polyclonal serum | 5–15 μg/mL | 1–10 μg/mL |
Crude ascites | 5–15 μg/mL | 1–10 μg/mL |
Affinity-purified polyclonal antibody | 1–12 μg/mL | 0.5–5 μg/mL |
Affinity-purified monoclonal antibody | 1–12 μg/mL | 0.5–5 μg/mL |
Recommended detection antibody concentration for ELISA in different systems. Check the user guide for the substrate as it may recommend a more defined concentration range for the enzyme conjugate.
Enzyme | System | Concentration |
---|---|---|
HRP | Colorimetric | 20–200 ng/mL |
Chemifluorescent | 25–50 ng/mL | |
Chemiluminescent | 10–100 ng/mL | |
AP | Colorimetric | 100–200 ng/mL |
Chemiluminescent | 40–200 ng/mL |
Using an enzyme-labeled secondary antibody
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
購買任何 ELISA 試劑組均可享有 折扣!
Purchase any Invitrogen ELISA kits at 25% off!/ 購買任何 ELISA 試劑組均可享有 25%的折扣!