The following package includes the GCOS Service Pack 2 software which updates GCOS client 1.3 and 1.4 workstations with a custom grid patch.
This patch includes:
- Components necessary to successfully grid image data (*.dat) and produce cell intensity data (*.cel) for the Genome-Wide Human SNP Array 5.0 and specific custom divide-by designs.
- This patch is applied to GCOS client systems only. It should not be installed on a GCOS server.
- This patch only affects arrays it was specifically designed for and is safe for systems running arrays outside of its scope.
Instructions for installing the update
- The ReadMe.txt file included in the download zip file contains detailed instructions for applying this update.
For Research Use Only. Not for use in diagnostic procedures.