Fluorescently labeled rat cerebellar neurons.
Filamentous structures of neuronal cells in a rat cerebellum were fluorescently labeled to differentiate the cell types. The cerebellum section was probed with primary antibodies to neurofilament and glial fibrillary acidic proteins (GFAP) and subsequently visualized with the green-fluorescent Alexa Fluor® 488 Goat Anti–Mouse IgG (Cat. No. A11001) and red-orange–fluorescent Alexa Fluor® 568 Goat Anti–Rabbit IgG (Cat. No. A11011) antibodies. This confocal micrograph was contributed by Gillian Davidson, Andrew Hubbard and Chris Guerin, Neurotoxicology Group, M.R.C Toxicology Unit, University of Leicester, Leicester, U.K.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Developing Drosophila embryo Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›