Developing Drosophila embryo
Formation of the cephalic furrow in the anterior end of a developing Drosophila melanogaster embryo visualized with the help of several fluorescent stains. A primary antibody to neurotactin was visualized using a red-fluorescent Cy3 dye secondary antibody (Amersham Pharmacia Biotech Ltd.). Primary antibodies to plasma membrane–bound myosin and to nuclear-localized even-skipped (Eve) protein were visualized with green-fluorescent Alexa Fluor® 488 Goat Anti–Mouse IgG antibody (Cat. No. A11001) and Alexa Fluor® 488 Goat Anti–Rabbit IgG antibody (Cat. No. A11008), respectively. The nuclei were stained with blue-fluorescent Hoechst 33342 (Cat. No. H1399, H3570, H21492). The sample was prepared by Eric Wieschaus, and the imaging was performed by Joe Goodhouse of Princeton University.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Endothelial cells. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›
Immunohistochemistry using GFAP Monoclonal Antibody, Mouse, Alexa Fluor® 594 Conjugate Go ›