">
Get Ready for the Future with Molecular Biology Innovations
Innovative products and workflow solutions
Advance your discoveries in science and set yourself up for future success. Check out the latest tools across molecular biology research applications that can enable fast and accurate results from sample to analysis.
Messaging Innovation to the Innovators
Jason Amsbaugh’s degree in biochemistry was
the starting point for a passion to communicate
with scientists. 
Build a custom cancer workflow to fit your research needs
From sample to analysis, explore end-to-end workflow solutions for liquid biopsy and solid tumor applications.
DNA & RNA isolation
Nucleic acid isolation is a critical step in many molecular biology applications. The integrity of isolated DNA and RNA is crucial for accurate and reliable downstream results. With the use of automated purification solutions that leverage magnetic bead-based isolation technology, high purity and yield can be achieved. Explore the latest in nucleic acid isolation that provides you with the reliability and versatility to enable a wide range of research applications. 
KingFisher Automated Purification Systems
Thermo Scientific KingFisher instruments provide easy-to-follow protocols that reduce overall prep time and errors associated with hands on sample purification. Save time, increase yield, and obtain reproducible results.
Sequential DNA/RNA isolation from hematologic samples
Enhance productivity and maximize precious hematologic cancer samples with sequential isolation of genomic DNA and total RNA. The Applied Biosystems MagMAX Sequential DNA/RNA Kit offers an automated workflow solution for total nucleic acid isolation from blood and bone marrow samples, enabling a more comprehensive downstream analysis.
KingFisher PlasmidPro Processor
Revolutionize your plasmid DNA extraction process for unparalleled efficiency and scalability with our cutting-edge innovation. The KingFisher PlasmidPro Maxi Processor introduces a breakthrough fully automated solution for maxi-scale purification.
Untouched CTC enrichment
Now with enhanced chemistry and performance, the Invitrogen Dynabeads MyOne CD45 Leukocyte Depletion beads offer untouched circulating tumor cell (CTC) enrichment through efficient depletion of human CD45-positive cells from whole blood, buffy coat, or peripheral blood mononuclear cells (PBMC).
Cancer research eBook
Explore the evolving field of cancer research through the lends of oncology biomarker applications
Protein isolation
The isolation of target proteins through the process of immunoprecipitation (IP) enables downstream analysis of their structure, function, and interactions. Workflows that begin with quality purification allow for the understanding of biological processes and disease mechanisms as well as drug discovery and development. Discover the potential of Dynabeads magnetic bead isolation technology that can provide the reproducibility you need to help achieve accurate and reliable downstream results. 
DynaGreen high-performing sustainable magnetic beads for immunoprecipitation
With more than 30 years of experience, the team behind Dynabeads has applied the 12 principles of green chemistry to create high-performing, non-microplastic magnetic beads that give reproducible results with low non-specific binding. From manufacture to customer site, Invitrogen DynaGreen magnetic beads were designed with sustainability in mind.
Dynabeads streptavidin for protein capture and beyond
Streptavidin-coupled Invitrogen Dynabeads magnetic beads can provide an exceptional platform for isolation and handling of biotinylated molecules and targets. From protein and cell capture to mRNA synthesis and target enrichment, this technology enables fast reproducible results for a wide variety of research applications.
Reverse transcription
Selecting the right reverse transcriptase is critical to detecting low-abundance RNAs in a sample and obtaining high yields of full-length cDNA. Historically, reverse transcription—the generation of complementary DNA from an RNA template—took over an hour with unpredictable yields. Now cDNA synthesis can be done in 10 minutes with more than 100x sensitivity.
Template switching reverse transcriptase kit
The Invitrogen SuperScript IV Template-Switching RT Master Mix contains a proprietary enhancer that maximizes template switching efficiency.
The latest in one-step RT-PCR
The Invitrogen SuperScript IV UniPrime One-Step RT-PCR System combines the power of SuperScript reverse transcriptase with universal primer annealing at 60°C to decrease optimization time and colored buffers for visual tracking of reaction setup and direct sample loading on gels, reducing pipetting errors.
Thermo Scientific EquiPhi29 DNA Amplification Kit
Enable fast, sensitive, and efficient isothermal amplification such as rolling circle amplification and multiple displacement amplification-whole genome amplification.
Amplification
Continued innovation allows Thermo Fisher Scientific to offer comprehensive nucleic acid amplification solutions designed for high performance, time savings, and reliability. In your pursuit to advance science, every experiment matters. There’s no time to start again and no need to wonder if the products you chose will set you back or propel you forward.
More sustainable PCR plastics
Thermo Scientific PCR Plastics, Sustain Series products are ISCC PLUS–certified, biobased PCR plastics represent our commitment to integrating more sustainable practices into our manufacturing processes to help reduce lab’s carbon footprint, without compromising on product quality or performance.
Ultimate flexibility and throughput
The Applied Biosystems ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems thermal cyclers with the flexible configuration and control features that fit how you work today. Interchangeable block formats allow you to maximize your throughput or run independent experiments concurrently.
Our latest in high-fidelity PCR enzymes
Invitrogen Platinum Superfi II DNA Polymerase is a hot-start, engineered proofreading DNA polymerase, providing exceptional fidelity and specificity to your PCR. With >300x Taq fidelity and buffer specially formulated for primer annealing at 60°C, Platinum SuperFi II DNA Polymerase offers efficiency and simplicity in PCR applications requiring high PCR accuracy, such as cloning, sequencing, and mutagenesis.
Discover the power of loop-mediated isothermal amplification
Invitrogen LAMP products are available in stand-alone or master mix formats, providing fast, highly sensitive, and specific LAMP reactions with resistance to a variety of inhibitors. They are also compatible with real-time and endpoint detection.
Cloning
Discover innovative products for the molecular cloning workflow. The molecular cloning workflow requires multiple steps to obtain high-quality cloned DNA for your downstream applications. Help advance your molecular cloning workflow at every step and accelerate construction of recombinant clones. Our wide range of cloning solutions address needs from routine to specialty applications, helping you find optimal reagents to achieve fast results with greater confidence.
One buffer for 176 enzymes—it's that easy
FastDigest restriction enzymes are an advanced line of 176 restriction enzymes in a complete, one-buffer system for simplified cloning. Performs complete digestion in 5–15 minutes and is compatible with downstream applications.
Easy everyday cloning
Invitrogen Gateway recombination cloning technology circumvents traditional restriction enzyme based cloning limitations, enabling you to access virtually any expression system in a few simple steps. From protein expression to functional analysis, Gateway cloning technology is applicable for a variety of research areas for multidisciplinary scientific studies.
The right tools for assembling multiple DNA fragments
GeneArt Seamless Cloning kits enable the simultaneous and directional assembly of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector for diverse applications.
Nucleic acid electrophoresis
Nucleic acid electrophoresis is a common technique in molecular biology to separate, identify, quantify, and/or purify nucleic acids. Traditionally, setting up electrophoresis involves numerous steps to achieve optimal separation and analysis of nucleic acid samples, such as gel preparation, ladder selection, sample visualization, and gel documentation. Now with the all-in-one, bufferless E-Gel Power Snap electrophoresis systems, save time and simplify your work—run, visualize, and image on one integrated device.
Separate DNA in as little as 10 minutes
Real-time visualization, flexible throughput, and easy analysis make the E-Gel Power Snap Plus system adaptable to any workflow. As the only system with the ability to run, visualize, and image all on a single, fully integrated device, the Power Snap Plus delivers high‑resolution images and the flexibility to run a few or up to 96 samples per run. 
Simply load samples and run
With E-Gel precast gels, no need to spend valuable time preparing gels. E-gels are self-contained and ready to use with agarose, electrodes, and DNA stains (Invitrogen SYBR Safe or SYBR Gold gel stains). They are packaged inside a disposable, UV-transparent cassette and can be used to analyze both DNA and RNA.
Safe analysis for you and your gels
Invitrogen SYBR Safe DNA Gel Stain was developed specifically to be less hazardous than ethidium bromide for staining DNA in agarose and acrylamide gels. It is a highly sensitive dye for visualizing DNA in agarose or acrylamide gels that can be viewed with blue-light or by UV excitation.
Flyer: Innovative molecular biology solutions
Enable your lab to be ready for the future with innovative molecular biology solutions across the workflow. Explore our latest products that can support your research success.
Brochure: KingFisher instruments
Prep for the future with KingFisher instruments. Automated purification for DNA, RNA, proteins, and cells.
KingFisher kit, reagent, and protocol selection tool
Find the right magnetic bead-based kit for your automated sample preparation needs.
Brochure: Dynabeads magnetic beads
Discover the power of Dynabeads magnetic bead technology and the range of applications Dynabeads products can support.
Molecular biology handbook
Unlock technical tips, educational tools, and guidance on selecting the right molecular biology products with our latest handbook.
Molecular Biology Personality Quiz
When working in the lab, understanding your unique research strengths can help you work more efficiently in the lab and help drive your innovations forward.
Accelerating the future of liquid biopsy brochure
Uncover the potential of liquid biopsy in oncology research and find product solutions from sample prep to data analysis.
Mol Bio Spotlight Articles
This collection features various molecular biology practical tips & tricks, break-though highlights, and the latest trends to help optimize your research.
Speaking of Mol Bio podcast
Listen to bold conversations that inspire confidence in the future of molecular biology to tackle complex, real-world problems.
Explore DNA & RNA Learning Center
Read scientific articles, webinars, and videos on nucleic acid research
School of Molecular Biology
Learn PCR and cloning at your own pace and on your own time.
For the Love of Innovation
Together, we make science exponentially better
For Research Use Only. Not for use in diagnostic procedures.
">