dataTaker loggers provide extensive features to be used within industry as a monitoring device in various applications.
With support for MODBUS sensors and SCADA communications, dataTaker loggers can be installed as standalone or integrated with an existing monitoring system.
DT82I | DT80 series | DT85 series |
---|---|---|
Low cost, low power data logger | Intermediate data logger | Advanced data logger |
IP54 | ||
- | Options for integrated modem | |
2 analog input channels | 5 analog input channels, expandable to 300 | 16 analog input channels, expandable to 960 |
4 digital input/counter channels | 8 digital input/counter channels | |
~10,000,000 data points | ||
- | USB port | |
2 serial ‘smart sensor’ ports | ||
Web and FTP client, RS232/485 | ||
- | SDI12 |
Increasing the channel capacity of the dataTaker DT80 and DT85 range is made very easy by adding dataTaker Channel Expansion Modules (CEM20). Each CEM20 connects 20 universal data logging channels to the dataTaker data logger.
dataTaker enclosures give you the flexibility to install your dataTaker system in a harsh or otherwise unsuitable environment by providing protection against external conditions such as water, dust, and accidental damage. These enclosures are suitable for all dataTaker systems and can be used for, industrial, weatherproof, or portable applications.