Reservoir engineering is a key discipline to increase and optimize hydrocarbon production. Reservoir engineers are responsible for designing scenarios for development of a reservoir. They build dynamic (multi-phase flow) models of the reservoir and perform economic evaluation of the asset. Reservoir engineers use Digital Rock Analysis (DRA) to calculate multi-phase flow properties on the rock samples from the reservoir that will help them in building the reservoir model.
PerGeos Software is a comprehensive Digital Rock Analysis solution that helps reservoir engineers build dynamic models of reservoirs and perform an evaluation of the asset.
PerGeos Software analyzes storage and flow capabilities of reservoir rocks based on imaging data. It also calculates multi-phase flow properties on the rock samples, including Relative Permeability, Capillary Pressure, Resistivity Index. Perform two-phase flow simulations.
Perform multi-scale geological and petrophysical analysis of your core imaging data
From Core to Pore, using imagery from large CT to FIB/SEM systems, PerGeos Software helps you digitally understand and validate observations and properties of your rock data with a high level of confidence.
Ability to interpret images at multiple resolutions from multiple modalities, advanced 2D/3D and 3D/3D registration tools and adaptive thresholding make PerGeos Software a unique digital platform for powerful up-scaling workflows.
This allows geological and petrophysical properties to be numerically obtained with confidence since all scales of the pore space can be analyzed and interpreted to calculate properties such as global porosity or relative permeability.